Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300175

Sigma-Aldrich

Monoclonal Anti-GSK3B antibody produced in mouse

clone 3D10, ascites fluid

Synonym(s):

GSK-3β, GSK3-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3D10, monoclonal

mol wt

46 kDa

species reactivity

human, mouse, monkey, rat

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSK3B(2932)

Immunogen

Purified recombinant fragment of human GSK3B expressed in E.coli.
Mouse monoclonal antibody raised against GSK3B

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madiha Kanwal et al.
Journal of Cancer, 15(11), 3466-3480 (2024-05-31)
Background: Cancer development involves alterations in key cellular pathways, with aspartate β-hydroxylase (ASPH) emerging as an important player in tumorigenesis. ASPH is upregulated in various cancer types, where it promotes cancer progression mainly by regulating the Notch1 and SRC pathways.
Ou Li et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 8510-8523 (2018-11-26)
BACKGROUND Long non-coding RNAs (lncRNAs) have been implicated in various human cancer types. However, the underlying mechanisms involved in hepatocellular carcinoma (HCC) progression remain poorly understood. MATERIAL AND METHODS In this study, lncRNA array was used to identify HCC related

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service