Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504676

Sigma-Aldrich

Anti-phospho-VE-Cadherin (pTyr731) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-7B4, Anti-CD144

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 87 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr731)

Gene Information

human ... CDH5(1003)

General description

Cadherin 5 (CDH5) is encoded by the gene mapped to long arm of human chromosome 16. The encoded protein is expressed in endothelial cells, but it is absent in normal epithelium.

Immunogen

The antiserum was produced against synthesized peptide derived from human VE-Cadherin around the phosphorylation site of Tyr731.

Immunogen Range: 697-746

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Cadherin 5 (CDH5) plays an essential role in various cellular processes, such as regulation of cell adhesion, cell proliferation, cell survival, cell shape, cell motility, modulation of signaling pathways and transcriptional gene regulation. Cdh5/VE(vascular endothelial)-cadherin is a core component of endothelial adherens junction, which plays a vital role in angiogenic sprouting by enhancing cell migration and cell elongation. CDH5 expressed in glioblastoma multiforme (GBM), facilitates the transdifferentiation of glioblastoma stem-like cells (GSCs) to endothelial cells and to form blood vessels, under hypoxia condition. VE-cadherin induces endothelial cell-cell adhesion at adherens junctions (AJs). CDH5 gene knockout results in loss of cell polarity and modification in vascular lumenal structure. CDH5 inhibits the synthesis of CDH2 (also known as N-cadherin) in the endothelium.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing-Gang Mao et al.
Neuro-oncology, 15(7), 865-879 (2013-05-07)
A proportion of glioblastoma stemlike cells (GSCs) expressing endothelial cell marker CDH5 (vascular-endothelial-cadherin or CD144) can transdifferentiate into endothelial cells and form blood vessels. However, the implications of CDH5 expression in gliomas and how it is regulated in GSCs remain
Frans van Roy
Nature reviews. Cancer, 14(2), 121-134 (2014-01-21)
Loss of cadherin 1 (CDH1; also known as epithelial cadherin (E-cadherin)) is used for the diagnosis and prognosis of epithelial cancers. However, it should not be ignored that the superfamily of transmembrane cadherin proteins encompasses more than 100 members in
Loïc Sauteur et al.
Cell reports, 9(2), 504-513 (2014-11-07)
Organ morphogenesis requires the coordination of cell behaviors. Here, we have analyzed dynamic endothelial cell behaviors underlying sprouting angiogenesis in vivo. Two different mechanisms contribute to sprout outgrowth: tip cells show strong migratory behavior, whereas extension of the stalk is dependent upon
Maria Grazia Lampugnani et al.
Journal of cell science, 123(Pt 7), 1073-1080 (2010-03-25)
Little is known about the molecular mechanisms that regulate the organization of vascular lumen. In this paper we show that lumen formation correlates with endothelial polarization. Adherens junctions (AJs) and VE-cadherin (VEC, encoded by CDH5) are required for endothelial apicobasal
Junwen Zhang et al.
Molecular medicine reports, 13(3), 2918-2924 (2016-02-06)
Changes in pulmonary microvascular permeability following cardiopulmonary bypass (CPB) and the underlying mechanisms have not yet been established. Therefore, the aim of the present study was to elucidate the alterations in pulmonary microvascular permeability following CPB and the underlying mechanism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service