Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504268

Sigma-Aldrich

Anti-phospho-TAK1 (pThr184) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pThr184)

Gene Information

human ... MAP3K7(6885)

General description

Transforming growth factor β (TGF-β)-activated kinase 1 (TAK1) is encoded by MAP3K7. TAK1 is a 606-residue protein, that contains a serine-threonine/tyrosine-protein kinase catalytic domain and a C-terminal coiled-coil domain. It is a member of the mitogen-activated protein kinase kinase kinase family (MAP3K). This gene is mapped to human chromosome 6q15.

Immunogen

The antiserum was produced against synthesized peptide derived from human TAK1 around the phosphorylation site of Thr184.

Immunogen Range: 161-210

Biochem/physiol Actions

Transforming growth factor β (TGF-β)-activated kinase 1 (TAK1) participates in the mitogen-activated protein kinase (MAPK)-p38 signaling pathway. This gene also participates in the autophagy process in retinal pigment epithelial cells. Mutations in MAP3K7 result in cardiospondylocarpofacial syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?III-tubulin overexpression is an independent predictor of prostate cancer progression tightly linked to ERG fusion status and PTEN deletion
Tsourlakis MC, et al.
The American Journal of Pathology, 184(3), 609-617 (2014)
TAK1 is involved in the autophagy process in retinal pigment epithelial cells.
Green YA
Biochemistry and Cell Biology = Biochimie Et Biologie Cellulaire, 94(2), 188-196 (2016)
Heterozygous Mutations in MAP3K7, Encoding TGF-?-Activated Kinase 1, Cause Cardiospondylocarpofacial Syndrome.
Le Goff C
American Journal of Human Genetics, 99(2), 407-413 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service