Skip to Content
Merck
All Photos(11)

Documents

SAB4503999

Sigma-Aldrich

Anti-phospho-TIE2 (pTyr1108) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD202B, Anti-GLC3E, Anti-TIE-2, Anti-VMCM, Anti-VMCM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 125 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1108)

Gene Information

human ... TEK(7010)

General description

The TIE2 (TEK receptor tyrosine kinase) is also termed as TEK (tyrosine-protein kinase receptor). This gene is located on human chromosome 9p21.2. It is expressed in the Schlemm′s canal endothelial of the eye. TIE2 belongs to the receptor tyrosine kinases family.

Immunogen

The antiserum was produced against synthesized peptide derived from human TIE2 around the phosphorylation site of Tyr1108.

Immunogen Range: 1074-1123

Biochem/physiol Actions

Tie-2 (TEK receptor tyrosine kinase) protein induces angiogenesis and supports endothelial cell survival, upon binding of angiopoietin-1 (an endothelial growth factor) to the receptor. This gene is necessary for Schlemm′s canal formation. This gene regulates vascular homeostasis by involving in auto or transphosphorylation. Tie-2 is essential for the growth of vascular structures during embryogenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Liu et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 54(10), e11028-e11028 (2021-07-22)
Engeletin is a natural derivative of Smilax glabra rhizomilax that exhibits anti-inflammatory activity and suppresses lipid peroxidation. In the present study, we sought to elucidate the mechanistic basis for the neuroprotective and pro-angiogenic activity of engeltin in a human umbilical
TIE2 Associates with Caveolae and Regulates Caveolin-1 To Promote Their Nuclear Translocation
Hossain MB, et al.
Molecular and Cellular Biology (2017)
Requisite role of angiopoietin-1, a ligand for the TIE2 receptor, during embryonic angiogenesis
Suri C, et al.
Cell, 87(7), 1171-1180 (1996)
Isolation of angiopoietin-1, a ligand for the TIE2 receptor, by secretion-trap expression cloning
Davis S, et al.
Cell, 87(7), 1161-1169 (1996)
Angiopoietin receptor TEK mutations underlie primary congenital glaucoma with variable expressivity.
Souma T
The Journal of Clinical Investigation, 126(7), 2575-2587 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service