Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503083

Sigma-Aldrich

Anti-Vimentin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

VIME, vimentin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VIM(7431)

General description

Anti-Vimentin Antibody detects endogenous levels of total Vimentin protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Vimentin.

Immunogen Range: 411-460

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunocytochemistry (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nurdan Özkucur et al.
Brain and behavior, 5(1), 24-38 (2015-02-28)
The disruption of neuron arrangement is associated with several pathologies. In contrast to action potentials, the role of resting potential (Vmem) in regulating connectivity remains unknown. Neuron assemblies were quantified when their Vmem was depolarized using ivermectin (Ivm), a drug
Richard A Lasher et al.
Journal of tissue engineering, 3(1), 2041731412455354-2041731412455354 (2012-08-25)
Quantifying structural features of native myocardium in engineered tissue is essential for creating functional tissue that can serve as a surrogate for in vitro testing or the eventual replacement of diseased or injured myocardium. We applied three-dimensional confocal imaging and
Ingrid Jurickova et al.
Inflammatory bowel diseases, 28(7), 988-1003 (2022-03-09)
Perturbagen analysis of Crohn's disease (CD) ileal gene expression data identified small molecules including eicosatetraynoic acid (ETYA), which may exert an antifibrotic effect. We developed a patient-specific human intestinal organoid (HIO) model system to test small molecule regulation of mitochondrial
Krizelle Mae M Alcantara et al.
Oncology reports, 41(4), 2103-2116 (2019-03-01)
Inactivation of the tumor suppressor protein Merlin leads to the development of benign nervous system tumors in neurofibromatosis type2 (NF2). Documented causes of Merlin inactivation include deleterious mutations in the encoding neurofibromin2 gene (NF2) and aberrant Merlin phosphorylation leading to
Heping Lin et al.
Oncology letters, 16(2), 1809-1814 (2018-07-17)
The long noncoding RNA (lncRNA) colon cancer-associated transcript 1 (CCAT1) has been identified as an oncogene in multiple types of human malignancy, and the aberrant expression of CCAT1 has been associated with the tumorigenesis and progression of cancer. However, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service