Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502631

Sigma-Aldrich

Anti-pan-Actin Antibody

rabbit polyclonal

Synonym(s):

ACTA, ACTA1, ACTS, Actin α skeletal muscle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Actin-pan antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTA1(58)

General description

Anti-Actin-pan antibody detects endogenous levels of total actin-pan protein.
The ACTA1 (actin, α1) gene is mapped to human chromosome 1q42.13. The gene codes for actin 1, a thin filament component of skeletal muscle.

Immunogen

The antiserum was produced against synthesized peptide derived from human Actin.

Immunogen Range: 21-70

Biochem/physiol Actions

Mutations in ACTA1 (actin, α1) is known to cause thin filament myopathy, such as nemaline myopathy, congenital myopathy, actin myopathy, intranuclear rod myopathy, cap myopathy and core myopathy. Upregulation of the gene is observed in orthotopic liver transplantation without any earlier evidence of autoimmune liver disease. The encoded protein actin forms a network of filaments and is responsible for the shape of the cell. It provides cellular strength and promotes active movement of the cell.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guosheng Luo et al.
Medical oncology (Northwood, London, England), 39(8), 111-111 (2022-06-07)
The ubiquitination process, which involves that binding of an ubiquitin protein to certain substrates, regulates several human biological processes and human cancers. Several studies report that the abnormal expression of quite a few E3 ubiquitin ligases could play critical role
Lack of Clinical Relevance of ANA and ASMA Positivity in Patients with Liver Transplantation without a History of Autoimmune Diseases.
Pellegrini L
BioMed Research International, 2017, 1-7 (2017)
Severe congenital actin related myopathy with myofibrillar myopathy features.
Selcen D
Neuromuscular Disorders, 25(6), 488-492 (2015)
Prevalence and phenotypes of congenital myopathy due to ?-actin 1 gene mutations.
Witting N
Muscle and Nerve, 53(3), 388-393 (2016)
Mutation-specific effects on thin filament length in thin filament myopathy.
Winter JM
Annals of Neurology, 79(6), 959-969 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service