Skip to Content
Merck
All Photos(2)

Key Documents

AB5585-I

Sigma-Aldrich

Anti-Recoverin Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Cancer-associated retinopathy protein, Cancer-associated retinopathy antigen, Protein CAR, Recoverin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rcvrn(19674)

General description

Recoverin (UniProt P35243; also known as Cancer-associated retinopathy antigen, Cancer-associated retinopathy protein, Protein CAR) is encoded by the RCVRN (also known as RCV1) gene (Gene ID 5957) in human. The photoreceptor protein recoverin is a member of the EF-hand neuronal calcium sensor (NCS) proteins. Recoverin is initially produced as a 200-amino acid protein whose N-terminal initiator methionine is removed posttranslationally. Recoverin is composed of four EF-hand domains (a.a. 25-60, 61-96, 97-132, 147-182), with the second and third EF-hand regions mediating low- and high-affinity calcium binding, respectively. In photoreceptor cells, recoverin functions as a Ca2+-dependent inhibitor of G-protein-coupled receptor kinase 1 (GRK1). Recoverin plays an important role in mediating calcium-dependent rod and cone light adaptation. Recoverin delays the rod & cone response under dim light when [Ca2+] is high by inhibiting GRK1-catalyzed visual pigment phosphorylation, but has little effect during light adaptation when [Ca2+]i is lowered.

Immunogen

Epitope: EF-hand 4
KLH-conjugated linear peptide corresponding to a sequence from the EF-hand 4 domain of human recoverin.

Application

Anti-Recoverin Antibody detects level of Recoverin and has been published and validated for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Recoverin in human and rat retina tissue sections.
Research Category
Neuroscience
Research Sub Category
Sensory & PNS

Quality

Evaluated by Western Blotting in human retina tissue lysate.

Western Blotting Analysis: 0.25 µg/mL of this antibody detected Recoverin in 10 µg of human retina tissue lysate.

Target description

~23 kDa observed.

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biju B Thomas et al.
Frontiers in neuroscience, 15, 752958-752958 (2021-11-13)
End-stage age-related macular degeneration (AMD) and retinitis pigmentosa (RP) are two major retinal degenerative (RD) conditions that result in irreversible vision loss. Permanent eye damage can also occur in battlefields or due to accidents. This suggests there is an unmet
Chien-Hui Lo et al.
The Journal of clinical investigation, 134(13) (2024-07-01)
Mitochondria-related neurodegenerative diseases have been implicated in the disruption of primary cilia function. Mutation in an intrinsic mitochondrial complex I component NDUFAF2 has been identified in Leigh syndrome, a severe inherited mitochondriopathy. Mutations in ARMC9, which encodes a basal body
Bin Lin et al.
Investigative ophthalmology & visual science, 61(11), 34-34 (2020-09-19)
To study if human embryonic stem cell-derived photoreceptors could survive and function without the support of retinal pigment epithelium (RPE) after transplantation into Royal College of Surgeons rats, a rat model of retinal degeneration caused by RPE dysfunction. CSC14 human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service