Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200820

Sigma-Aldrich

Anti-Fetuin A antibody, Mouse monoclonal

clone FetA-M, purified from hybridoma cell culture

Synonym(s):

Alpha-2-HS-glycoprotein (AHSG), Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
FetA-M, monoclonal
application:
species reactivity:
human
technique(s):
immunoblotting: 2-4 μg/mL using Human serum
citations:
4

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

FetA-M, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using Human serum

isotype

IgM

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AHSG(197)

General description

Fetuin A, also known as Alpha-2-HS-glycoprotein (AHSG), Alpha-2-Z-globulin or Ba-alpha-2-glycoprotein, is a member of the cystatin superfamily of cysteine protease inhibitors. Fetuin A is a secreted calcium-regulatory glycoprotein produced by liver Hepatocytes. It is cleaved into two chains bound by disulfide bonds and inhibits ectopic Ca×PO4 ion precipitation and vascular calcification.
Monoclonal Anti-Fetuin A (mouse IgM isotype) is derived from the Fet A-M hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a BALB/C mouse immunized with a synthetic peptide corresponding to the internal region of human AHSG (Fetuin A), conjugated to KLH as immunogen. Fetuin A is a 64 kDa protein, predominantly localized to the blood.

Specificity

Monoclonal Anti-Fetuin A specifically recognizes Fetuin A from human plasma.

Immunogen

Synthetic peptide corresponding to human Fution A internal region, conjugated to KLH

Application

The antibody is recommended for use in various immunological techniques including Immunoblotting, Immunohistochemistry and ELISA.

Biochem/physiol Actions

Fetuin A is a secreted calcium-regulatory glycoprotein produced by liver hepatocytes. It is cleaved into two chains bound by disulphide bonds and inhibits ectopic Ca×PO4 ion precipitation and vascular calcification.
Reduction in the serum fetuin might indicate coronary artery disease. Fetuin is also implicated in insulin resistance and metabolic syndrome. Increased levels of fetuin A is observed in non-alcoholic fatty liver disease.
Fetuin A is consider to be a negative acute-phase reactant and its serum concentration decreases in response to inflammation, compared with normal levels. In addition, it acts as a mineral chaperone to facilitate the transport and removal of potentially proinflammatory and procalcific cargo (waste).Fetuin A plays a major role in numerous diseases and pathophysiological conditions involved with calcium metabolisms.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apoptosis-mediated endothelial toxicity but not direct calcification or functional changes in anti-calcification proteins defines pathogenic effects of calcium phosphate bions
Kutikhin AG, et al.
Scientific Reports, 6, 27255-27255 (2016)
The relationship of circulating fetuin-a with liver histology and biomarkers of systemic inflammation in nondiabetic subjects with nonalcoholic fatty liver disease
Celebi G, et al.
Saudi Journal of Gastroenterology : Official Journal of the Saudi Gastroenterology Association, 21(3), 139-139 (2015)
Emerging human Fetuin A assays for biomedical diagnostics
Vashist SK, et al.
Trends in biotechnology, 35(5), 407-421 (2017)
Chelsea Miller et al.
Journal of biomedical materials research. Part A, 111(8), 1096-1109 (2023-01-03)
Surface modifications can be applied to biomaterials to alter the various surface properties that influence protein-material interactions and the cellular response. The plasma protein fetuin-A has been found to adsorb to many biomaterials but details of its interactions with polydimethylsiloxane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service