Skip to Content
Merck
All Photos(4)

Key Documents

U4375

Sigma-Aldrich

Uridine 5′-diphospho-N-acetylglucosamine sodium salt

≥98%

Synonym(s):

UDP-GlcNAc, UDP-N-acetylglucosamine, UDPAG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C17H25N3O17P2Na2
CAS Number:
Molecular Weight:
651.32
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51
Pricing and availability is not currently available.

biological source

bakers yeast

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

[Na+].[Na+].CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@@H]1OP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](O)[C@@H]2O)N3C=CC(=O)NC3=O

InChI

1S/C17H27N3O17P2.2Na/c1-6(22)18-10-13(26)11(24)7(4-21)35-16(10)36-39(31,32)37-38(29,30)33-5-8-12(25)14(27)15(34-8)20-3-2-9(23)19-17(20)28;;/h2-3,7-8,10-16,21,24-27H,4-5H2,1H3,(H,18,22)(H,29,30)(H,31,32)(H,19,23,28);;/q;2*+1/p-2/t7-,8-,10-,11-,12-,13-,14-,15-,16-;;/m1../s1

InChI key

HXWKMJZFIJNGES-YZVFIFBQSA-L

General description

Uridine 5′-diphospho-N-acetylglucosamine (UDP-GlcNAc) is a nucleotide sugar. It is synthesized from glucose via the hexosamine biosynthetic pathway (HBP).[1] UDP-GlcNAc is transported actively into Golgi, nucleotide sugar transporter (NST).[2] The levels of UDP-GlcNAc is modulated by the concentration of nutrients exposed to the cell.[3]

Application

Uridine 5′-diphospho-N-acetylglucosamine (UDP-GlcNAc) sodium salt has been used:
  • as a component of reaction cocktail in endoplasmic reticulum to Golgi transport assay[4]
  • as a reference standard for the quantification of UDP-GlcNAc in liver tissues using high-performance liquid chromatography (HPLC)[5]
  • in testing the glycosylation activity of O-GlcNAc transferase (OGT) against peptide substrate[6]

Biochem/physiol Actions

Donor substrate for transfer of GlcNAc to the dolichol precursor in the synthesis of N-glycans. UDP-GlcNAc also functions as the source of GlcNAc attached to serine/threonine residues by O-GlcNAc transferase (OGT) in O-GlcNAc signaling.
Uridine 5′-diphospho-N-acetylglucosamine (UDP-GlcNAc) is a sugar donor and aids in the endomembrane glycosylation of endoplasmic reticulum and Golgi. The decreased levels of UDP-GlcNAc has an influence on the normal cellular proliferation and apoptosis.[1] UDP-GlcNAc elicits feedback inhibition of the enzyme glutamine:fructose-6-phosphate amidotransferase (GFAT).[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Golgi UDP-GlcNAc transporter delivers substrates for N-linked glycans and sphingolipids
Ebert B, et al.
Nature plants, 1-1 (2018)
Microsome-Based Assay for Analysis of Endoplasmic Reticulum to Golgi Transport in Mammalian Cells
Cell Biology, 209-214 (2006)
Katsuhiro Sawasato et al.
Scientific reports, 9(1), 1372-1372 (2019-02-06)
MPIase is a glycolipid that is involved in membrane protein integration. Despite evaluation of its functions in vitro, the lack of information on MPIase biosynthesis hampered verification of its involvement in vivo. In this study, we found that depletion of
A little sugar goes a long way: the cell biology of O-GlcNAc
Bond MR and Hanover JA
The Journal of cell biology, 208(7), 869-880 (2015)
LXRalpha Regulates Hepatic ChREBPalpha Activity and Lipogenesis upon Glucose, but Not Fructose Feeding in Mice
Fan Q, et al.
Nutrients, 9(7), 678-678 (2017)

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

LC-MS/MS method quantifies similar polar nucleotide activated sugars using Supel™ Carbon LC column for simultaneous analysis.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service