Skip to Content
Merck
All Photos(1)

Documents

SAB3701282

Sigma-Aldrich

Anti-Human IgG (Fc specific)-Peroxidase antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

HRP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Human IgG coupled to agarose beads. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Peroxidase, Anti-Rabbit Serum, Human IgG, Human IgG F(c) and Human Serum. No reaction was observed against Human IgG F(ab′)2

Immunogen

Human IgG F(c) fragment

Physical properties

Antibody format: IgG

Physical form

Supplied in 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Reconstitution

Reconstitute with 1.0 mL deionized water (or equivalent).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda S Machado et al.
Parasitology, 147(9), 932-939 (2020-04-21)
The co-infection between visceral leishmaniasis (VL) and human immunodeficiency virus (HIV) has increased in several countries in the world. The current serological tests are not suitable since they present low sensitivity to detect the most of VL/HIV cases, and a
Renata G F Alvim et al.
Biochemical engineering journal, 186, 108537-108537 (2022-07-26)
Serological tests detect antibodies generated by infection or vaccination, and are indispensable tools along different phases of a pandemic, from early monitoring of pathogen spread up to seroepidemiological studies supporting immunization policies. This work discusses the development of an accurate
Graziele Fonseca de Sousa et al.
PeerJ, 10, e14547-e14547 (2022-12-22)
To analyze the long-term dynamics of antibodies against SARS-CoV-2 and understand the impact of age, gender, and viral load on patients' immunological response. Serum samples were obtained from 231 COVID-19 positive patients from Macaé, in Rio de Janeiro state, in
Graziele F Sousa et al.
Viruses, 15(7) (2023-07-29)
COVID-19 has infected humans worldwide, causing millions of deaths or prolonged symptoms in survivors. The transient or persistent symptoms after SARS-CoV-2 infection have been defined as post-COVID-19 conditions (PCC). We conducted a study of 151 Brazilian PCC patients to analyze
Beatriz C S Salles et al.
Immunobiology, 224(1), 163-171 (2018-09-30)
Serological tests are important tools for the diagnosis of Leishmania infection. However, they are not effective markers to diagnose asymptomatic cases of visceral leishmaniasis (VL) and patients developing tegumentary leishmaniasis (TL), since antileishmanial antibodies can be encountered in low levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service