Skip to Content
Merck
All Photos(2)

Key Documents

SAB3500816

Sigma-Aldrich

Anti-CMPK2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 49 kDa

species reactivity

mouse, rat, human

technique(s)

ELISA: suitable
immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CMPK2(129607)

General description

Cytidine/uridine monophosphate kinase 2 (CMPK2) is expressed in mitochondria. The protein Cmpk2 is similar to thymidylate kinase than to cytosolic Cmpk1. Cmpk2 has thymidylate kinase domain and the C-terminal domain contains all the consensus motifs: P-loop (ATP/ GTP binding motif A), LID (domain in lipase) domain, adenine-base binding loop, catalytic site, and potential substrate binding site. In human chromosome, the gene is localized on 2p25.2.

Biochem/physiol Actions

Cmpk2 utilizes ATP (adenosine triphosphate) as phosphate donor to phosphorylate dUMP (deoxyuridine monophosphate), dCMP (deoxycytidine monophosphate), CMP (cytosine monophosphate), and UMP (uridine monophosphate). The order of substrate specificity for Cmpk2 is UMP, the highest, followed by dCMP, CMP and UMP. Cmpk2 also phosphorylates monophosphate nucleoside analogs ddC (zalcitabine), dFdC (gemcitabine), araC (vidarabine), BVDU (brivudine), and FdUrd (fluorodeoxyuridine). Cmpk2 expression is found in leukemic cells like chronic myelogenous leukemia K-562 and lymphoblastic leukemia MOLT-4, implicates its pathophysiology. The catalytic activity of Cmpk2 is similar to human adenylate kinase 9.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500816.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human UMP-CMP kinase 2, a novel nucleoside monophosphate kinase localized in mitochondria
Xu Y, et al.
The Journal of Biological Chemistry, 283(3), 1563-1571 (2008)
Marjan Amiri et al.
The international journal of biochemistry & cell biology, 45(5), 925-931 (2013-02-19)
Adenylate kinases regulate adenine nucleotide levels and are present in different intracellular compartments. These enzymes also participate in the activation of pharmacologically active nucleoside and nucleotide analogs. We have in the present study identified the ninth isoform of the adenylate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service