Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900181

Sigma-Aldrich

Anti-FAP (AB1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

170-kDa melanoma membrane-bound gelatinase, FAP, FAP alpha, FAPA, Seprase, fibroblast activation protein, fibroblast activation protein alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 10-20 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FAP(2191)

Related Categories

General description

FAP gene is localized on human chromosome 2q24.2.
Fibroblast activation protein (FAP) is a type II transmembrane serine protease and a cell surface antigen. It is present as a homodimeric integral protein with dipeptidyl peptidase IV like fold. FAP has an α/β-hydrolase domain and an eight-bladed β-propeller domain. It is not expressed in normal tissues. The protein is only expressed by activated fibroblasts in response to pathologic situations.

Immunogen

Synthetic 18 amino acid peptide from extracellular domain of human FAP. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (100%); Monkey (94%); Bovine (89%); Marmoset (83%).

Application

Anti-FAP (AB1) antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Fibroblast activation protein (FAP) is expressed in several pathogenic sites including cancer, fibrosis, arthritis, wounding, or inflammation. FAP has in vitro dipeptidyl peptidase activity and collagenolytic activity. It cleaves N-terminal dipeptides from polypeptides and can degrade gelatin and type I collagen.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Xuguang Yang et al.
Cancer research, 76(14), 4124-4135 (2016-05-25)
Cancer-associated fibroblasts (CAF) are components of the tumor microenvironment whose contributions to malignant progression are not fully understood. Here, we show that the fibroblast activation protein (FAP) triggers induction of a CAF subset with an inflammatory phenotype directed by STAT3
L Wagner et al.
Clinical and experimental immunology, 184(3), 265-283 (2015-12-17)
Dipeptidyl peptidase (DPP) 4 (CD26, DPP4) is a multi-functional protein involved in T cell activation by co-stimulation via its association with adenosine deaminase (ADA), caveolin-1, CARMA-1, CD45, mannose-6-phosphate/insulin growth factor-II receptor (M6P/IGFII-R) and C-X-C motif receptor 4 (CXC-R4). The proline-specific
Fibroblast activation protein-a: a key modulator of the microenvironment in multiple pathologies
Thomas Kelly-
International Review of Cell and Molecular Biology, 297 (2012)
Ellen Puré et al.
Oncogene, 37(32), 4343-4357 (2018-05-04)
Fibroblast activation protein (FAP) is a cell-surface serine protease that acts on various hormones and extracellular matrix components. FAP is highly upregulated in a wide variety of cancers, and is often used as a marker for pro-tumorigenic stroma. It has
Fibroblast Activation Protein, a Dual Specificity Serine Protease Expressed in Reactive Human Tumor Stromal Fibroblasts*
John E Park
The Journal of Biological Chemistry (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service