Skip to Content
Merck
All Photos(8)

Documents

P0372

Sigma-Aldrich

Anti-Podocin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Podocin Antibody, Podocin Antibody - Anti-Podocin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~42 kDa (doublet)

species reactivity

human, rat, mouse

technique(s)

indirect immunofluorescence: 10-20 μg/mL using acetone-fixed human or rat kidney frozen sections
western blot (chemiluminescent): 0.5-1 μg/mL using whole extract of rat glomeruli

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NPHS2(7827)
mouse ... Nphs2(170484)
rat ... Nphs2(170672)

Related Categories

General description

Podocin is a podocyte structural protein, that belongs to the band-7-stomatin family of lipid raft-associated proteins. This family contains more than 1800 conserved proteins which share ∼150 amino acid domains showing similarity to mitochondrial protein prohibitin (PHB).
Podocin is a hairpin-like integral membrane protein with intracellular N- and C- termini. Podocin is located at the insertion site of the slit membrane, an intercellular junction found in mammalian kidney.

Specificity

Anti-Podocin recognizes human, mouse, and rat podocin.

Immunogen

Synthetic peptide corresponding to amino acid residues of human podocin with an N-terminal added cysteine, conjugated to KLH. The corresponding sequence is identical in mouse and differs by one amino acid in rat.

Application

Anti-Podocin antibody produced in rabbit has been used for Western Blotting and immunohistochemistry.

Biochem/physiol Actions

Podocin and nephrin are essential for maintaining the slit diaphragm. Studies show that mutations in this gene account for around 15% cases of nephrotic syndrome cases. Podocin interacts with cholesterol and promotes the formation of protein-lipid supercomplexes, which control ion channel complexes. The podocin gene is known to be associated with autosomal recessive SRNS (nephrotic syndrome type 2). Podocin actively participates in the ultrafiltration process occuring in kidney.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month.
For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NPHS2 mutations account for only 15% of nephrotic syndrome cases.
Guaragna MS et al
BMC Medical Genetics, 16(88) (2015)
Excess podocyte semaphorin-3A leads to glomerular disease involving plexinA1-nephrin interaction.
Reidy KJ et al
The American Journal of Pathology, 183(4), 1156-1168 (2013)
Induction of podocyte VEGF164 overexpression at different stages of development causes congenital nephrosis or steroid-resistant nephrotic syndrome.
Veron D et al
The American Journal of Pathology, 177(5), 2225-2233 (2010)
The SPFH domain-containing proteins: more than lipid raft markers.
Browman D T, et al.
Trends in Cell Biology, 17(8), 394-402 (2007)
Batoul Wehbi et al.
Journal of the American Society of Nephrology : JASN, 30(7), 1238-1249 (2019-06-23)
IgA nephropathy (IgAN) often follows infections and features IgA mesangial deposition. Polymeric IgA deposits in the mesangium seem to have varied pathogenic potential, but understanding their pathogenicity remains a challenge. Most mesangial IgA1 in human IgAN has a hypogalactosylated hinge

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service