Skip to Content
Merck
All Photos(2)

Documents

SAB2106550

Sigma-Aldrich

Anti-MYH7 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CMD1S, Anti-CMH1, Anti-MPD1, Anti-MYHCB, Anti-SPMD, Anti-SPMM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

223 kDa

species reactivity

guinea pig, bovine, horse, rabbit, mouse, human, rat, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYH7(4625)
mouse ... Myh7(140781)

Immunogen

Synthetic peptide directed towards the middle region of human MYH7

Biochem/physiol Actions

Myh7 play a role in muscle contraction.

Sequence

Synthetic peptide located within the following region: TLEDQMNEHRSKAEETQRSVNDLTSQRAKLQTENGELSRQLDEKEALISQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Ren et al.
Cell death discovery, 7(1), 378-378 (2021-12-09)
Cardiac hypertrophy is a common pathological change accompanied by various cardiovascular diseases; however, its underlying mechanisms remain elusive. Mounting evidence indicates that long non-coding RNAs (lncRNAs) are novel transcripts involved in regulating multiple biological processes. However, little is known about
Hanping Qi et al.
Molecular therapy. Nucleic acids, 19, 507-522 (2020-01-11)
Cardiac hypertrophy, a response of the heart to increased workload, is a major risk factor for heart failure. Myostatin (MSTN) is an inhibitor of myogenesis, regulating the number and size of skeletal myocytes. In recent years, cardiomyocyte autophagy also has
Kevin M Tharp et al.
Cell metabolism, 27(3), 602-615 (2018-03-08)
The activation of brown/beige adipose tissue (BAT) metabolism and the induction of uncoupling protein 1 (UCP1) expression are essential for BAT-based strategies to improve metabolic homeostasis. Here, we demonstrate that BAT utilizes actomyosin machinery to generate tensional responses following adrenergic
Diego Salagre et al.
Antioxidants (Basel, Switzerland), 12(8) (2023-08-26)
Obesity-induced skeletal muscle (SKM) inflexibility is closely linked to mitochondrial dysfunction. The present study aimed to evaluate the effects of melatonin on the red vastus lateralis (RVL) muscle in obese rat models at the molecular and morphological levels. Five-week-old male
Yapeng Li et al.
Experimental cell research, 370(1), 78-86 (2018-06-15)
Metabolic dysfunction is a hallmark of cardiac hypertrophy and heart failure. During cardiac failure, the metabolism of cardiomyocyte switches from fatty acid oxidation to glycolysis. However, the roles of key metabolic enzymes in cardiac hypertrophy are not understood fully. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service