Merck
  • Actomyosin-Mediated Tension Orchestrates Uncoupled Respiration in Adipose Tissues.
Actomyosin-Mediated Tension Orchestrates Uncoupled Respiration in Adipose Tissues.
Cell metabolism (2018-03-08)
Kevin M Tharp, Michael S Kang, Greg A Timblin, Jon Dempersmier, Garret E Dempsey, Peter-James H Zushin, Jaime Benavides, Catherine Choi, Catherine X Li, Amit K Jha, Shingo Kajimura, Kevin E Healy, Hei Sook Sul, Kaoru Saijo, Sanjay Kumar, Andreas Stahl
ABSTRACT
The activation of brown/beige adipose tissue (BAT) metabolism and the induction of uncoupling protein 1 (UCP1) expression are essential for BAT-based strategies to improve metabolic homeostasis. Here, we demonstrate that BAT utilizes actomyosin machinery to generate tensional responses following adrenergic stimulation, similar to muscle tissues. The activation of actomyosin mechanics is critical for the acute induction of oxidative metabolism and uncoupled respiration in UCP1+ adipocytes. Moreover, we show that actomyosin-mediated elasticity regulates the thermogenic capacity of adipocytes via the mechanosensitive transcriptional co-activators YAP and TAZ, which are indispensable for normal BAT function. These biomechanical signaling mechanisms may inform future strategies to promote the expansion and activation of brown/beige adipocytes.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Anti-MYH7 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Blebbistatin, Racemic, InSolution, ≥97%, 50 mM in 90% DMSO, reversible inhibitor of nonmuscle myosin II
Sigma-Aldrich
(−)-Isoproterenol hydrochloride
Sigma-Aldrich
Anti-UCP-1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Latrunculin B, Latrunculia magnifica, Latrunculin B, CAS 76343-94-7, is a unique marine toxin that inhibits actin polymerization and disrupts microfilament organization. It is 10 to 100-fold more potent than cytochalasins.
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.
Sigma-Aldrich
Oligomycin, A mixture of A, B, and C isomers.
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder