Skip to Content
Merck
All Photos(1)

Key Documents

S178

Sigma-Aldrich

(R)-SKF-82957 hydrobromide

solid

Synonym(s):

(1R)-6-Chloro-2,3,4,5-tetrahydro-3-methyl-1-phenyl-1H-3-benzazepine-7,8-diol hydrobromide, 1H-3-Benzazepine-7,8-diol,6-chloro-2,3,4,5-tetrahydro-3-methyl-1-phenyl hydrobromide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18ClNO2 · HBr
CAS Number:
Molecular Weight:
384.70
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

optical activity

[α]22/D +22.46°, c = 0.76 in DMF(lit.)

color

white

solubility

H2O: 7.5 mg/mL
DMSO: soluble

SMILES string

Br[H].CN1CCc2c(Cl)c(O)c(O)cc2[C@H](C1)c3ccccc3

InChI

1S/C17H18ClNO2.BrH/c1-19-8-7-12-13(9-15(20)17(21)16(12)18)14(10-19)11-5-3-2-4-6-11;/h2-6,9,14,20-21H,7-8,10H2,1H3;1H/t14-;/m1./s1

InChI key

BCZHWHQKKPEORG-PFEQFJNWSA-N

Biochem/physiol Actions

Selective D1 dopamine receptor agonist.

Caution

hygroscopic, photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P H Andersen et al.
European journal of pharmacology, 188(6), 335-347 (1990-06-12)
Dopamine receptor selectivity was investigated for a number of dopamine receptor agonists. In vitro, the benzazepine derivatives, e.g., SKF 38393 and SKF 75670 as well as the isoquinoline derivatives, SKF 89626 and SKF 89615, were D1 receptor-selective. All other compounds
F R Pfeiffer et al.
Journal of medicinal chemistry, 25(4), 352-358 (1982-04-01)
6-Chloro-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepines were synthesized and evaluated as agonists of central and peripheral dopamine receptors. These benzazepines were prepared by cyclization of certain amino alcohols followed by demethylation of the 7,8-dimethoxy groups of the precursors to the 7,8-catecholic moiety. Preliminary evidence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service