Skip to Content
Merck
All Photos(1)

Key Documents

S1172

Sigma-Aldrich

Anti-Syntaxin 1 antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Synonym(s):

Anti-GEFSP9, Anti-STX1B1, Anti-STX1B2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB CL
clone:
polyclonal
species reactivity:
mouse, rat
citations:
19
technique(s):
immunohistochemistry: suitable using rat brain sections
western blot (chemiluminescent): 1:1,000 using rat brain membranes

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse, rat

technique(s)

immunohistochemistry: suitable using rat brain sections
western blot (chemiluminescent): 1:1,000 using rat brain membranes

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

Syntaxin 1 is a membrane protein. It is also known as HPC-1. Neurons express syntaxin 1A.

Immunogen

highly purified GST fusion protein with the residues 1-265 of rat syntaxin 1A (intracellular, N-terminal).

Application

Anti-Syntaxin 1 antibody has been used in western blotting and immunohistochemistry.

Biochem/physiol Actions

Syntaxin 1 (STX1A) may control exocytosis in synaptic vesicles. In retina, STX1A may help in the structural formation of the INL (inner nuclear layer) and OPL(outer plexiform layer). Deletion of STX1A in mice affects the functions of hypothalamic-pituitary-adrenal axis. It may participate in spinal nociceptive plasticity promoted by peripheral nerve injury.

Physical form

After reconstitution, the antibody solution is phosphate buffered saline, pH 7.4, containing 1% BSA and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic exercise enhances insulin secretion ability of pancreatic islets without change in insulin content in non-diabetic rats
Tsuchiya M, et al.
Biochemical and Biophysical Research Communications, 430(2), 676-682 (2013)
Impaired NGF/TrkA Signaling Causes Early AD-Linked Presynaptic Dysfunction in Cholinergic Primary Neurons
Latina V, et al.
Frontiers in Cellular Neuroscience, 11 (2017)
Tiziana Cotrufo et al.
The European journal of neuroscience, 36(9), 3152-3164 (2012-09-06)
Directed cell migration and axonal guidance are essential steps in neural development that share many molecular mechanisms. The guidance of developing axons and migrating neurons is likely to depend on the precise control of plasmalemma turnover in selected regions of
SNARE protein expression in synaptic terminals and astrocytes in the adult hippocampus: a comparative analysis
Schubert V, et al.
Glia, 59(10), 1472-1488 (2011)
A Critical role for Bim in retinal ganglion cell death
McKernan DP and Cotter TG
Journal of Neurochemistry, 102(3), 922-930 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service