Skip to Content

RAB1936

Human MIS/AMH ELISA

for Serum, Plasma, and Cell Culture Supernatants

Synonym(s):

AMH, Anti-Muellerian hormone, MIS, Muellerian-inhibiting factor, Muellerian-inhibiting substance

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Gene information:
human ... AMH(268)
Input:
sample type serum
sample type cell culture supernatant(s)
sample type plasma
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.18 ng/mL
standard curve range: 0.164-40 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AMH(268)

General description

This ELISA antibody pair detects Human MIS. Other species not determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.


Kit Components Only

Product No.
Description

  • RAB1936A HUMAN MIS ELISA PLATE (ITEM A) 96 wells

  • RAB1936C LYOPHILIZED HUMAN MIA PROTEIN STANDARD (ITEM C) 2 vials

  • RAB1936F BIOTINYLATED HUMAN MIA DETECTION ANTIBODY (ITEM F) 2 vials

Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • 20X Wash Buffer (Item B) 25 mL
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H) 12 mL
    SDS

  • ELISA Stop Solution (Item I) 8 mL
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer E (Item E2) 15 mL
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library