Skip to Content
Merck
All Photos(1)

Key Documents

RAB1038

Sigma-Aldrich

Human ECM-1 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum sample(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 24 pg/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ECM1(1893)

General description

Extracellular matrix protein 1 (ECM1) is a glycoprotein. It is expressed in skin and other tissues. The gene encoding this protein is localized on human chromosome 1q21.2.
This ELISA antibody pair detects Human Extracellular Matrix Protein 1 (ECM1)

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Extracellular matrix protein 1 (ECM1) has a role in cell proliferation, mineralization and angiogenesis. The protein has been shown to have a role in cancer invasion and progression. It is associated with hepatocellular carcinoma. Mutations in the ECM1 gene have been linked to lipoid proteinosis. The protein also promotes the Warburg effect, which is an oncogenic switch that allows cancerous cells to take in more glucose than non-cancerous cells.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alee Choi et al.
International journal of molecular sciences, 21(17) (2020-09-03)
Extracellular matrix (ECM) components play an important role in maintaining skeletal muscle function, but excessive accumulation of ECM components interferes with skeletal muscle regeneration after injury, eventually inducing fibrosis. Increased oxidative stress level caused by dystrophin deficiency is a key
Hege Kampen Pihlstrøm et al.
PloS one, 15(12), e0243759-e0243759 (2020-12-17)
Following a successful renal transplantation circulating markers of inflammation may remain elevated, and systemic inflammation is associated with worse clinical outcome in renal transplant recipients (RTRs). Vitamin D-receptor (VDR) activation is postulated to modulate inflammation and endothelial function. We aimed
Kyung-Eun Lee et al.
International journal of molecular sciences, 21(22) (2020-11-14)
In recent years, a number of active materials have been developed to provide anti-aging benefits for skin and, among them, peptides have been considered the most promising candidate due to their remarkable and long-lasting anti-wrinkle activity. Recent studies have begun
Cristina Bárcena et al.
EBioMedicine, 43, 513-524 (2019-05-12)
Chronic hepatic inflammation leads to liver fibrosis, which may progress to cirrhosis, a condition with high morbidity. Our aim was to assess the as yet unknown role of innate immunity protein CD5L in liver fibrosis. CD5L was measured by ELISA
Extracellular matrix protein 1 gene (ECM1) mutations in nine Iranian families with lipoid proteinosis
Izadi F, et al.
The Indian Journal of Medical Research, 143(3), 303-303 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service