Skip to Content
Merck
All Photos(1)

Key Documents

RAB0742

Sigma-Aldrich

Human TPO / Thyroid Peroxidase ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type urine
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.3 ng/mL
standard curve range: 0.41-100 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TPO(7173)

General description

The TPO ELISA kit provides for the Quantitative measurement of Thyroid Peroxidase in Cell Culture Supernatants, Plasma and Serum.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADDELISA 5X Assay/Sample Diluent Buffer D (Item K)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Walter Jackson et al.
Cancer research, 77(8), 1942-1954 (2017-02-17)
Little is known about how megakaryocytes may affect metastasis beyond serving as a source of platelets. In this study, we explored the functional implications of megakaryocyte accumulation in the femurs of mice after injection of metastatic or non-metastatic breast cancer
Kuniaki Ota et al.
Human reproduction (Oxford, England), 35(6), 1276-1287 (2020-06-02)
Is there any relationship between vitamin D [25 (OH) vitamin D], total plasma homocysteine and methylenetetrahydrofolate reductase (MTHFR) C677T polymorphism in women with recurrent pregnancy losses (RPL)? Women with MTHFR 677TT (homozygous mutation, TT) genotype have significantly lower vitamin D
Mei Han et al.
Bioorganic & medicinal chemistry, 27(12), 2480-2486 (2019-03-07)
To explore whether Arctigenin-Valine ester (ARG-V) can treat myelosuppression caused by chemotherapy. The number of peripheral blood cells of the mice was measured by an automatic blood analyzer, and the hematopoietic progenitor colonies CFU-GM, CFU-E, BFU-E, and CFU-Meg were cultured
Elisa Tinazzi et al.
Autoimmunity reviews, 19(9), 102616-102616 (2020-07-20)
Immune Thrombocitopenic Purpura (ITP) is an autoimmune disease characterized by antibody-mediated platelet destruction and variable reduced platelet production. Besides antibody-mediated platelet destruction, new pathogenic mechanisms have been reported to be involved in reducing platelet count. Among these, desialylation is one
Liang Li et al.
Molecular medicine reports, 23(1) (2020-11-13)
Huangqi, the dried root of Radix Astragali, is an essential herb in Traditional Chinese Medicine and has been used to promote hematopoiesis for centuries. Astragalus polysaccharide (ASPS), the bioactive compound of Huangqi, serves a crucial role in hematopoiesis. The aim of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service