Skip to Content
Merck

RAB0564

Mouse Tnfsf4 / Tumor Necrosis Factor Ligand Superfamily Member 4 ELISA Kit

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Gene information:
mouse ... Tnfsf4(22164)
Input:
sample type serum
sample type plasma
sample type cell culture supernatant(s)
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 30 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Tnfsf4(22164)

General description

This ELISA antibody pair detects mouse TNFSF4/OX40.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

Tumor necrosis factor (TNF) superfamily member 4 (Tnfsf4) signals specifically through the OX40 receptor, which is expressed primarily on CD4+T cells but also on certain activated CD8+T cells. Tnfsf4 acts as a costimulatory signal, which is essential for a productive interaction between antigen presenting cells and their target T-cells. It enhances cell proliferation and survival and increases expression of regulated on activation, normal T cell expressed and secreted (RANTES) or chemokine ligand 5 (CCL5), interleukins-2 and -3 and interferon γ (IFNg). Tnfsf4 signaling plays an important role in immuno-regulatory communication and immune-tolerance. Polymorphisms in the TNFSF4 gene leads to the development of autoimmune thyroid diseases (AITDs), rheumatoid arthritis (RA), systemic lupus erythematosus (SLE) and systemic sclerosis (SSc).

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library