RAB0546
Human LAG3 / Lymphocyte Activation Gene 3 Protein ELISA Kit
for serum, plasma, cell culture supernatants and urine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type cell culture supernatant(s)
sample type urine
sample type plasma
sample type serum
assay range
inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 45 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... LAG3(3902)
General description
This ELISA antibody pair detects human Lymphocyte Activation Gene 3 Protein.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lawrence P Andrews et al.
Science immunology, 5(49) (2020-07-19)
Mechanisms of resistance to cancer immunotherapy remain poorly understood. Lymphocyte activation gene-3 (LAG3) signaling is regulated by a disintegrin and metalloprotease domain-containing protein-10 (ADAM10)- and ADAM17-mediated cell surface shedding. Here, we show that mice expressing a metalloprotease-resistant, noncleavable LAG3 mutant
Yingying Sun et al.
International journal of hematology, 112(6), 757-763 (2020-08-14)
To investigate the expression of lymphocyte activation gene 3(LAG3) on CD8+T effector cells (Teffs), CD4+ Teffs and regulatory T cells (Tregs) in patients with severe aplastic anemia (SAA). We detected the expression of LAG3 on CD8+ Teffs, CD4+ Teffs and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service