Skip to Content
Merck
All Photos(1)

Key Documents

RAB0520

Sigma-Aldrich

Bovine IFNG / Interferon Gamma ELISA Kit

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

bovine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.11 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... IFNG(281237)

General description

This ELISA antibody pair detects bovine Interferon Gamma.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janyra A Espinoza et al.
Journal of immunology (Baltimore, Md. : 1950), 199(1), 212-223 (2017-06-02)
Human respiratory syncytial virus (hRSV) is the leading cause of severe lower respiratory tract infections in children. The development of novel prophylactic and therapeutic antiviral drugs against hRSV is imperative to control the burden of disease in the susceptible population.
Kawaljit Kaur et al.
Oncoimmunology, 7(5), e1426518-e1426518 (2018-05-04)
Therapeutic role of NK cells in solid tumors was challenged previously even though their role in hematological malignancies has clearly been established. Furthermore, functions and numbers of NK cells are greatly suppressed in oral cancer patients necessitating effective future NK
Vanessa A Evans et al.
AIDS (London, England), 32(11), 1491-1497 (2018-05-11)
In HIV-infected individuals on antiretroviral therapy (ART), latent HIV is enriched in CD4 T cells expressing immune checkpoint molecules, in particular programmed cell death-1 (PD-1). We therefore assessed the effect of blocking PD-1 on latency, both in vitro and in
Marcelo Augusto Kazuo Ikeda et al.
Frontiers in microbiology, 9, 2286-2286 (2018-10-20)
Sporotrichosis is a mycosis that affects the skin, lymphatic system and other organs in humans and animals. The disease has a worldwide distribution, with endemic areas in Brazil, and is caused by a complex of species, including Sporothrix brasiliensis. Some
Ryosuke Uchiyama et al.
Journal of immunology (Baltimore, Md. : 1950), 199(3), 1122-1130 (2017-07-05)
CD4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service