Skip to Content
Merck

RAB0472

Human TIMP-2 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

Metalloproteinase inhibitor 2 , TIMP-2, Tissue inhibitor of metalloproteinases 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... TIMP2(7077)
Input:
sample type urine
sample type cell culture supernatant(s)
sample type serum
sample type plasma
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type urine
sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 2 pg/mL
standard curve range: 1.6-1200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TIMP2(7077)

General description

The Human TIMP-2 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human TIMP-2 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human TIMP2

Application

For research use only. Not for use in diagnostic procedures.
Human TIMP-2 ELISA Kit has been used to measure tissue inhibitor of metalloproteinase-2 (TIMP-2) level in blood samples from Atherosclerotic renal artery stenosis (ARAS) patients.

Biochem/physiol Actions

Tissue inhibitor of metalloproteinase-2 (TIMP-2) plays a vital role in inhibition of matrix metalloproteinases (MMPs) and functions as a key modulator of extracellular matrix deposition and degradation in various physiological processes. TIMP-2 can also act through a matrix metalloproteinase (MMP)-independent mechanism inhibiting endothelial cell proliferation in vitro. TIMP-2 mediates anti-angiogenic activities in vivo. TIMP-2 is a target gene of the microRNA-miR-22. TIMP-2 functions as a biomarker for cell cycle arrest.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library