Skip to Content
Merck

RAB0375

Human Carbohydrate Antigen 15-3 / Mucin-1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

CA 15-3, Cancer antigen 15-3, EMA, MUC-1, Mucin-1 , Tumor-associated epithelial membrane antigen, CA15-3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... MUC1(4582)
Input:
sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type plasma
sample type urine
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 4 mU/mL
standard curve range: 4.1-1000 mU/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MUC1(4582)

General description

The Human CA15-3 (Cancer Antigen) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CA15-3 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Human CA15-3

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 1X Assay/Sample Diluent Buffer A (Item D1)
    SDS

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Khayal Al-Khayal et al.
Oncology letters, 12(1), 393-400 (2016-06-28)
Mucin overexpression has been implicated in the tumorigenesis and progression of colorectal carcinoma (CRC). However, data obtained on the prognostic importance of mucin expression in CRC is inconsistent. Due to lack of data on mucin expression and the increase in
Tianhe Huang et al.
International journal of molecular sciences, 16(2), 2956-2970 (2015-02-03)
Human epididymis protein 4 (HE4) is a recognized biomarker in ovarian and endometrial cancer and over-expressed in pancreatic adenocarcinoma. The diagnostic value of HE4 in pancreatic adenocarcinoma remains unknown. Here we elucidate mRNA, protein and serum level of HE4 in



Global Trade Item Number

SKUGTIN
RAB0375-1KT04061834962069