Skip to Content
Merck
All Photos(2)

Key Documents

RAB0312

Sigma-Aldrich

Rat Interleukin-6 ELISA Kit

for for cell lysate and tissue lysate

Synonym(s):

IL-6, Interleukin-6 

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type tissue lysate
sample type cell lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il6(24498)

General description

The Rat IL-6 ELSA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IL-6 cell lysate and tissue lysate.

Immunogen

Recombinant Rat IL-6

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Rat Interleukin-6 ELISA Kit has been used to determine the plasma level of interleukin-6 (IL-6) via ELISA.[1]

Biochem/physiol Actions

Interleukin-6 (IL-6) acts as a neuromodulation factor. IL-6 has two signalling pathways, known as classic signalling pathway and trans-signalling pathway. It has a neuroprotective role in ischemic stroke, by activating IL-6/interleukin 6 receptor (IL-6R)/signal transducers and activators of transcription 3 (STAT3) signalling pathway.[2] IL-6 is a multifunctional cytokine, which is implicated in acute inflammation.[3] IL-6 is associated with SAPHO (synovitis, acne, pustulosis, hyperostosis, and osteitis) syndrome.[4]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interleukin-6 RNA knockdown ameliorates acute lung injury induced by intestinal ischemia reperfusion in rats by upregulating interleukin-10 expression
Yuan B, et al.
Molecular Medicine Reports, 16(3), 2529-2537 (2017)
AT1 receptor blocker losartan protects against mechanical ventilation-induced diaphragmatic dysfunction
Kwon OS, et al.
Journal of Applied Physiology, 119(10), 1033-1041 (2015)
Effects of interleukin-6 and IL-6/AMPK signaling pathway on mitochondrial biogenesis and astrocytes viability under experimental septic condition
Chen XL, et al.
International Immunopharmacology, 59, 287-294 (2018)
Thomas A Rasmussen et al.
The lancet. HIV, 5(5), e221-e230 (2018-04-13)
Whether ongoing virus replication occurs in HIV-infected individuals on antiretroviral therapy (ART) is unclear; therefore, whether residual virus replication is a barrier to achieving a cure for HIV is also unknown. We aimed to establish whether ART intensification with dolutegravir
Mike Recher et al.
NPJ vaccines, 3, 17-17 (2018-05-26)
Pneumococcal conjugate vaccine (PCV) is recommended for adults with chronic disease. Extensive limb swelling (ELS) is an acute vigorous local inflammatory reaction following vaccination. Predisposing immune system correlates and the influence of ELS on vaccine responses are not known. Here

Questions

  1. Good morning i'm a PhD sudent WE have bought a kit of rat interleukin 6 but WE don't fond the prospectus in the box So please if it's possible to send me the prospectus Cordially

    1 answer
    1. The protocol for this kit is lot specific and is provided in the Certificate of Analysis. See the link below for an example:
      https://www.sigmaaldrich.com/certificates/Graphics/COfAInfo/SigmaABNO/RAB0312_1128J0733.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service