Skip to Content
Merck
All Photos(2)

Key Documents

RAB0299

Sigma-Aldrich

Mouse IL-4 ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.6 pg/mL
standard curve range: 0.82-200 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il4(16189)

General description

The Mouse IL-4 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IL-4 in serum, plasma and cell culture supernatants.

Immunogen

Recombinant Mouse IL-4

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing-Ge Zhang et al.
International immunopharmacology, 20(2), 307-315 (2014-04-08)
Cholecystokinin octapeptide (CCK-8), an immunomodulatory peptide, can promote or suppress the development or function of specific CD4(+) T cell subsets by regulating antigen-presenting cell functions. In the current study, we investigated whether CCK-8 exerts a direct effect on T cells
Christine Hartoonian et al.
Molecular biology reports, 41(9), 5943-5952 (2014-06-29)
Plasmid DNA vaccination is a promising vaccine platform for prevention and treatment of infectious disease. Enhancement of the DNA vaccine potency by co-inoculation of immunoadjuvant has been shown to be an effective strategy. Modulation of dendritic cells and T-cells locomotion
Fen He et al.
Journal of neuroimmunology, 271(1-2), 1-7 (2014-04-10)
Patients received cranial-irradiation can be affected with cognitive deficits and decreasing hippocampal neurogenesis. In this work, we characterized the cognitive ability and immune-induced neurogenesis of the pre- and post-treated cranial-irradiated rats with Glatiramer acetate (GA), known as a weak CNS
Mohamed El-Behi et al.
European journal of immunology, 44(10), 3003-3014 (2014-07-30)
IL-17-secreting CD8(+) T cells (Tc17 cells) have been implicated in immunity to infections, cancer, and autoimmune diseases. Thus far, studies on Tc17 cells have primarily investigated their development from naïve precursors, while the biology of committed Tc17 cells has been
Marwa Abdallah et al.
Experimental dermatology, 23(10), 742-747 (2014-08-12)
Not only macrophages, T-helper (Th)1 and Th2, but also CD4(+) CD25(high) FoxP3(+) regulatory T cells (T-regs) are involved in immune response to Mycobacterium leprae. We aimed to evaluate serum interleukin (IL)-1β and IL-12p70 (macrophage cytokines), interferon-γ (IFN-γ) (Th1 cytokine), IL-4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service