Skip to Content

RAB0108

Mouse CT-1 ELISA Kit

for serum, plasma and cell culture supernatant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
mouse ... Ctf1(12977)
Input:
sample type serum
sample type plasma
sample type cell culture supernatant(s)
Species reactivity:
mouse
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 200 pg/mL
standard curve range: 0.246-60 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ctf1(12977)

General description

The Mouse CT-1 (Cardiotrophin-1) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Mouse CT-1 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse Cardiotrophin-1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 5X Assay/Sample Diluent Buffer B (Item E1)
    SDS

  • ELISA 1X Assay/Sample Diluent Buffer C (Item L)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library