Skip to Content
Merck
All Photos(1)

Key Documents

R8626

Sigma-Aldrich

Rifamycin SV sodium salt

potency: ≥ 900 IU/mg (anhydrous basis)

Synonym(s):

2,7-(Epoxypentadeca(1,11,13)trienimino)naphtho(2,1-b)furan-1,11(2H)-dione, 5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl- 21-acetate monosodium salt, Rifamycin SV monosodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H46NNaO12
CAS Number:
Molecular Weight:
719.75
EC Number:
UNSPSC Code:
51283601
PubChem Substance ID:
NACRES:
NA.76

form

powder

Quality Level

potency

≥ 900 IU/mg (anhydrous basis)

solubility

ethanol: soluble 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
mycobacteria

Mode of action

protein synthesis | interferes

SMILES string

[Na+].CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)cc([O-])c4c3C2=O

InChI

1S/C37H47NO12.Na/c1-16-11-10-12-17(2)36(46)38-23-15-24(40)26-27(32(23)44)31(43)21(6)34-28(26)35(45)37(8,50-34)48-14-13-25(47-9)18(3)33(49-22(7)39)20(5)30(42)19(4)29(16)41;/h10-16,18-20,25,29-30,33,40-44H,1-9H3,(H,38,46);/q;+1/p-1/b11-10+,14-13+,17-12-;/t16-,18+,19+,20+,25-,29-,30+,33+,37-;/m0./s1

InChI key

YVOFSHPIJOYKSH-NLYBMVFSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Rifamycin SV is a broad-spectrum semisynthetic antimicrobial agent of the rifamycin group. It is used to study potential genes, such as rif15 and rif16, which may be involved in the conversion of rifamycin SV into rifamycin B.

Biochem/physiol Actions

Rifamycin SV inhibits selective (E. coli, B. subtilis) bacterial DNA-dependent RNA polymerase by binding to the polymerase β-subunit, a mechanism similar to rifabutin. It acts as a selective cytochrome P450 3A4 inducer. It is active against Gram-positive bacteria and is moderately active against Gram-negative organisms.

Packaging

1g,5g

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Air sensitive. Store under inert gas

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Kono et al.
Journal of biochemistry, 91(1), 397-401 (1982-01-01)
The formation of Mn2+- or Cu2+-rifamycin SV complex and the semiquinone radical of rifamycin SV was studied by electron spin resonance spectroscopy. The results indicate that: (1) the complexes Mn2+-rifamycin SV (g parallel = 2.265, g perpendicular = 2.087, A
I Caruso et al.
Annals of the rheumatic diseases, 41(3), 232-236 (1982-06-01)
Thirty rheumatoid patients with persistent knee effusion were treated intra-articularly with rifamycin SV, 500 mg weekly, or with saline solution, 10 ml, in a double-blind study. A complete disappearance of effusion and an impressive clinical improvement was observed in the
Marta Calatayud et al.
Chemical research in toxicology, 25(2), 446-453 (2012-01-05)
Inorganic arsenic (iAs) [As(III)+As(V)] is a drinking water contaminant, and human exposure to these arsenic species has been linked with a wide range of health effects. The main path of exposure is the oral route, and the intestinal epithelium is
Eric Garver et al.
Drug metabolism and disposition: the biological fate of chemicals, 36(12), 2492-2498 (2008-08-30)
Macrolide antibiotics azithromycin (AZI) and clarithromycin (CLARI) are large molecular weight compounds and are substrates for apically polarized efflux transporters such as P-glycoprotein, which can potentially restrict intestinal absorption. However, despite these undesired physicochemical and biopharmaceutical properties, AZI and CLARI
E Vigliaroli et al.
Minerva stomatologica, 59(9), 507-517 (2010-10-14)
Actinomycosis is an uncommon chronic granulomatous infection that cause formation of abscesses and cutaneous fistula. In mandibular actinomycosis the alveolar bone and mandibular body are usually not involved and the pathogenetic mechanisms of the actinomycotic infiltration is unknown. The patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service