Skip to Content
Merck
All Photos(1)

Documents

R3257

Sigma-Aldrich

Resorufin sodium salt

redox indicator

Synonym(s):

7-Hydroxy-3H-phenoxazin-3-one sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H6NNaO3
CAS Number:
Molecular Weight:
235.17
Beilstein:
174850
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

form

powder

mp

>300 °C (lit.)

solubility

water: 1 mg/mL, opaque

storage temp.

room temp

SMILES string

[Na+].[O-]c1ccc2N=C3C=CC(=O)C=C3Oc2c1

InChI

1S/C12H7NO3.Na/c14-7-1-3-9-11(5-7)16-12-6-8(15)2-4-10(12)13-9;/h1-6,14H;/q;+1/p-1

InChI key

TYXBZZBYMSVFPR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Resorufin is a fluorescent indicator.

Application

Resorufin has been used to determine the cytochrome P450 (CYP) 2A5 coumarin 7-hydroxylase (COH) activity changes over a broad range of lipopolysaccharide (LPS) doses. It has also been used as an internal standard for ethoxyresurufin-o-deethylase (EROD) analysis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of biological effects of environmental pollution in Mersin Bay (Turkey, northeastern Mediterranean Sea) using Mullus barbatus and Liza ramada as target organisms
Yilmaz D, et al.
Environmental Pollution (Barking, Essex : 1987), 208, 361-370 (2016)
Tomas Hansson et al.
Archives of environmental contamination and toxicology, 66(2), 237-247 (2013-12-04)
Since the new millennium, a notion has developed in certain parts of society that environmental pollutants and their associated effects are under control. The primary objective of this investigation, performed in 2003, was to test whether this was actually the
L Wang et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 28(3), 215-227 (2000-06-14)
Hepatocytes encapsulated in alginate-poly-1-lysine-alginate (APA) are used in transplantation studies and in bioartificial liver support systems. Loss of cell viability in the process of APA encapsulation is usually 20-30% while the effect on cytochrome CYP450 activity is rarely reported. This
Alessandro Trentini et al.
Clinical chemistry and laboratory medicine, 58(10), 1749-1758 (2020-02-08)
Background Myeloperoxidase (MPO) is an enzyme with a recognized prognostic role in coronary artery disease (CAD), which is also emerging as a promising biomarker for cardiac risk stratification. However, the lack of a consensus method for its quantification has hindered
2013 IEEE International Conference on Solid Dielectrics (ICSD) IEEE (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service