Skip to Content
Merck
All Photos(1)

Documents

28605

Sigma-Aldrich

3-Cyanoumbelliferone

BioReagent, suitable for fluorescence, ≥98.0% (TLC)

Synonym(s):

3-Cyano-7-hydroxycoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5NO3
CAS Number:
Molecular Weight:
187.15
Beilstein:
153271
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.32

product line

BioReagent

Assay

≥98.0% (TLC)

form

powder

mp

≥250 °C (lit.)

solubility

DMF: soluble
alcohols: soluble

fluorescence

λex 408 nm; λem 450 nm in methanol

suitability

suitable for fluorescence

SMILES string

Oc1ccc2C=C(C#N)C(=O)Oc2c1

InChI

1S/C10H5NO3/c11-5-7-3-6-1-2-8(12)4-9(6)14-10(7)13/h1-4,12H

InChI key

IJQYTHQDUDCJEQ-UHFFFAOYSA-N

Gene Information

human ... MIF(4282)

Application

3-Cyano-7-hydroxycoumarin is closely related to 3-cyano-7-ethoxycoumarin which is used as a fluorometric substrate and inhibitor for cytochrome P-450 enzymes and cytochrome P-450-dependent mixed function oxidases. 3-Cyano-7-hydroxycoumarin may be useful to study the kinetics and substrate specificity of cytochrome P-450s.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay J Agarwal et al.
PloS one, 8(5), e63028-e63028 (2013-05-15)
TRAM-34, a clotrimazole analog characterized as a potent and selective inhibitor of intermediate-conductance, calcium-activated K(+) (IKCa) channels, has been used extensively in vitro and in vivo to study the biological roles of these channels. The major advantage of TRAM-34 over
Joonyoung F Joung et al.
Physical chemistry chemical physics : PCCP, 19(37), 25509-25517 (2017-09-14)
Proton dissociation (PD) reactions of weak acids and proton transfer (PT) processes in aqueous solutions are strongly influenced by ions. However, a detailed molecular picture that describes how ions affect the rates of PD and PT processes is still missing.
Samuel Koenig et al.
Aquatic toxicology (Amsterdam, Netherlands), 108, 11-17 (2011-11-19)
Variations in cytochrome P450 enzyme (CYPs) distribution and function between animal groups could result in differential metabolism and elimination kinetics for certain contaminants. Although a number of studies have suggested that differences in polychlorobiphenyl (PCB) accumulation profiles between crustacea and
Ilana Berger et al.
PloS one, 6(11), e26794-e26794 (2011-11-10)
Cytosolic sulfotransferases (SULTs) are mammalian enzymes that detoxify a wide variety of chemicals through the addition of a sulfate group. Despite extensive research, the molecular basis for the broad specificity of SULTs is still not understood. Here, structural, protein engineering
Corinna Krempl et al.
Insect biochemistry and molecular biology, 78, 69-77 (2016-10-18)
Gossypol is a polyphenolic secondary metabolite produced by cotton plants, which is toxic to many organisms. Gossypol's aldehyde groups are especially reactive, forming Schiff bases with amino acids of proteins and cross-linking them, inhibiting enzyme activities and contributing to toxicity.

Articles

Nitric oxide (NO) as a signal transporter in neurons, endothelial cells and in the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service