Skip to Content
Merck
All Photos(1)

Key Documents

R1003

Sigma-Aldrich

Ribonuclease T1 from Aspergillus oryzae

ammonium sulfate suspension, 300,000-600,000 units/mg protein

Synonym(s):

Guanyloribonuclease, Ribonucleate 3′-guanylo-oligonucleotidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

Aspergillus sp. (Aspergillus oryzae)

form

ammonium sulfate suspension

specific activity

300,000-600,000 units/mg protein

mol wt

11068 by amino acid sequence

technique(s)

cell based assay: suitable

suitability

suitable for separating native or denatured proteins, or nucleic acids

application(s)

cell analysis

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Ribonuclease T1 (RNase T1) from Aspergillus oryzae is used to digest denatured RNA prior to sequencing and is used for protein folding studies .

Biochem/physiol Actions

Ribonuclease T1 (RNase T1) from Aspergillus oryzae is an endoribonuclease that hydrolyzes after G residues. Cleavage occurs between the 3′-phosphate group of a guanidine ribonucleotide and 5′-hydroxyl of the adjacent nucleotide. The initial product is a 2′:3′ cyclic phosphate nucleoside that is hydrolyzed to the corresponding 3′-nucleoside phosphate. It differs from Pancreatic RNase in that it attacks the guanine sites specifically to yield 3′-GMP and oligonucleotides with a 3′-GMP terminal group.

Unit Definition

One unit will produce acid soluble oligonucleotides equivalent to a ΔA260 of 1.0 in 15 min at pH 7.5 at 37°C, in a reaction volume of 1.0 mL. Substrate: Yeast RNA.

Physical form

Suspension in 2.8 M (NH4)2SO4 solution

Analysis Note

Protein determined by E1%/280

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ribonuclease T1: Structure, Function, and Stability
Pace, CN; Heinemann U, et al.
Angewandte Chemie (International Edition in English), 4, 343-360 (1991)
Catherine Brands et al.
Journal of visualized experiments : JoVE, (160) (2020-07-07)
In situ hybridization is a powerful technique to identify specific RNA or DNA sequences within individual cells in tissue sections, providing important insights into physiological processes and disease pathogenesis. In situ hybridization (ISH) has been used for many years to
Elisa Bombarda et al.
The journal of physical chemistry. B, 114(5), 1994-2003 (2010-01-22)
Because of their central importance for understanding enzymatic mechanisms, pK(a) values are of great interest in biochemical research. It is common practice to determine pK(a) values of amino acid residues in proteins from NMR or FTIR titration curves by determining
Rasmus Lybech Jensen et al.
Journal of the American Chemical Society, 134(23), 9820-9826 (2012-05-19)
Singlet molecular oxygen, O(2)(a(1)Δ(g)), can influence many processes pertinent to the function of biological systems, including events that result in cell death. Many of these processes involve a reaction between singlet oxygen and a given amino acid in a protein.
Scott Quarrier et al.
RNA (New York, N.Y.), 16(6), 1108-1117 (2010-04-24)
Structure mapping experiments (using probes such as dimethyl sulfate [DMS], kethoxal, and T1 and V1 RNases) are used to determine the secondary structures of RNA molecules. The process is iterative, combining the results of several probes with constrained minimum free-energy

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service