Skip to Content
Merck
All Photos(1)

Key Documents

10109134001

Roche

RNase

from bovine pancreas

Synonym(s):

Rnase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204

biological source

bovine pancreas

Quality Level

form

dry powder (prepared from crystallized ribonuclease)

specific activity

~40 U/mg dry powder (dry powder according to Kunitz (+25°C; RNA as substrate).)

packaging

pkg of 500 mg

manufacturer/tradename

Roche

shipped in

wet ice

storage temp.

2-8°C

General description

Crude mixture of RNases.Dry powder (prepared from crystallized ribonuclease).
Human chromosome 14q11.2 shows the presence of all human RNases. Only eight (RNase 1-8) of them were true called as canonical RNases. And the rest nine to thirteen are called as noncanonical RNases. Human RNase 1 is called as HP-RNase or human pancreatic RNase. Human RNase 2 is also called as EDN (eosinophils derived neurotoxin) that is isolated from eosinophils.

Specificity

Heat inactivation: 1 mM EDTA

Application

RNase has been used in DNA ladder assay.
RNase has been used in analysis of DNA fragmentation using agarose gel electrophoresis. It has been used during flow cytometry analysis.

Unit Definition

One unit is the enzyme activity that causes a decrease in absorbance of A0 to A1 within 1 minute under the assay conditions; A0 to A1 corresponds to the total conversion, A1 being the final absorbance.

Preparation Note

Activator: PK requires Mg2+ (or Mn2+, Co2+) and K+ (or NH4+, Rb+) for full activity.
Working solution: It is recommended to dissolve the RNase (1 mg/ml) in water or TE buffer, pH 7.5.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ribonucleases (2011)
Ognjen Sekulović et al.
Methods in molecular biology (Clifton, N.J.), 1476, 143-165 (2016-08-11)
Bacteriophages (phages) are present in almost, if not all ecosystems. Some of these bacterial viruses are present as latent "prophages," either integrated within the chromosome of their host, or as episomal DNAs. Since prophages are ubiquitous throughout the bacterial world
S Saito et al.
The Journal of biological chemistry, 275(44), 34528-34533 (2000-08-10)
Apoptosis of cardiac myocytes is one of the causes of heart failure. Here we examine the mechanism by which the activation of beta-adrenergic receptor induces cardiomyocyte apoptosis. Terminal deoxynucleotide transferase-mediated dUTP nick end labeling and DNA ladder analyses revealed that
David Olmeda et al.
Molecular biology of the cell, 14(7), 2844-2860 (2003-07-15)
Beta-catenin is a multifunctional protein involved in cell-cell adhesion and Wnt signal transduction. Beta-catenin signaling has been proposed to act as inducer of cell proliferation in different tumors. However, in some developmental contexts and cell systems beta-catenin also acts as
Mihnea R Mangalea et al.
Cell host & microbe, 29(5), 726-739 (2021-05-07)
Rheumatoid arthritis (RA) is an autoimmune disease characterized in seropositive individuals by the presence of anti-cyclic citrullinated protein (CCP) antibodies. RA is linked to the intestinal microbiota, yet the association of microbes with CCP serology and their contribution to RA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service