Skip to Content
Merck
All Photos(1)

Key Documents

PZ0378

Sigma-Aldrich

CP-809,101 fumarate

≥98% (HPLC)

Synonym(s):

2-(3-Chlorobenzyloxy)-6-(piperazin-1-yl)pyrazine fumarate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H17ClNO4 · C4H4O4
CAS Number:
Molecular Weight:
426.82
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

storage temp.

−20°C

SMILES string

Clc1cc(ccc1)COc2nc(cnc2)N3CCNCC3

InChI

1S/C15H17ClN4O/c16-13-3-1-2-12(8-13)11-21-15-10-18-9-14(19-15)20-6-4-17-5-7-20/h1-3,8-10,17H,4-7,11H2

InChI key

PCWGGOVOEWHPMG-UHFFFAOYSA-N

Biochem/physiol Actions

CP-809,101 is a potent and high-affinity 5-hydroxytryptamine receptor 2C (5-HT2C) full agonist with only weaker partial agonist activity toward 5-HT2A & 5-HT2B (cellular Ca2+ mobilization EC50 in nM/efficiency = 0.11/93% and 0.06/97% with human and rat 5-HT2C, respectively; 153/67%/human 5HT2A, 65.3/57%/human 5HT2B, 119/79%/rat 5HT2A) and much reduced or little affinity toward other receptors, ion channels and uptake sites tested. CP-809,101 exhibits antipsychotic efficacy in reducing conditioned avoidance response/CAR (ED50 = 4.8 mg/kg s.c.) and locomotor activity (ED50 in mg/kg via s.c. = 2/rats, 1/mice; no effect up to 10 mg/kg in 5-HT2C-knockout mice), while oral administration is reported to suppress spontaneous and nocturnal food intake as well as fasting-induced refeeding in rats (∼8% and ∼24% reduction of cumulative food intake and body weight in 4 days, respectively; 30 mg/kg/day p.o.).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amit S Kalgutkar et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(6), 848-858 (2007-03-09)
2-(3-Chlorobenzyloxy)-6-(piperazin-1-yl)pyrazine (3) is a potent and selective 5-HT(2C) agonist that exhibits dose-dependent inhibition of food intake and reduction in body weight in rats, making it an attractive candidate for treatment of obesity. However, examination of the genotoxicity potential of 3
Judith A Siuciak et al.
Neuropharmacology, 52(2), 279-290 (2006-09-05)
CP-809,101 is a potent, functionally selective 5-HT(2C) agonist that displays approximately 100% efficacy in vitro. The aim of the present studies was to assess the efficacy of a selective 5-HT(2C) agonist in animal models predictive of antipsychotic-like efficacy and side-effect
Paul J Fletcher et al.
Neuropharmacology, 57(3), 259-267 (2009-06-09)
5-HT(2C) receptor agonists have considerable therapeutic potential, however there is little in vivo data to compare the potency and selectivity of 5-HT(2C) receptor agonists. Since 5-HT(2C) receptor agonists reduce locomotor activity and food intake, changes in these drug-induced behaviours in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service