Skip to Content
Merck
All Photos(1)

Key Documents

PZ0124

Sigma-Aldrich

CP-66713

≥98% (HPLC)

Synonym(s):

8-chloro-1-phenyl-[1,2,4]triazolo[4,3-a]quinoxalin-4-amine, PF-1716311

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClN5
CAS Number:
Molecular Weight:
295.73
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

Nc1nc2ccc(Cl)cc2n3c(nnc13)-c4ccccc4

InChI

1S/C15H10ClN5/c16-10-6-7-11-12(8-10)21-14(9-4-2-1-3-5-9)19-20-15(21)13(17)18-11/h1-8H,(H2,17,18)

InChI key

PBENJWAFQLORQL-UHFFFAOYSA-N

Biochem/physiol Actions

CP-66713 is an adenosine A2 receptor antagonist.

Features and Benefits

This compound is featured on the Adenosine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M O'Kane et al.
European journal of pharmacology, 362(1), 17-25 (1998-12-29)
Previous work has been carried out on the effects of adenosine on transmitter release and on the excitability of postsynaptic neurones, but little is known about the effects of adenosine on the coupling between the two. In this study, we
S Fujii et al.
Neuroscience letters, 148(1-2), 148-150 (1992-12-14)
The effects of the adenosine A2 receptor antagonist CP-66713 on the reversal of long-term potentiation (LTP) were studied in CA1 neurons of guinea pig hippocampal slices. Reduction of LTP (depotentiation, DP) was effected by delivering a train of low-frequency afferent
T Matsugi et al.
Investigative ophthalmology & visual science, 38(13), 2695-2701 (1998-01-07)
To investigate the effect of adenosine on the contractile tone of cultured bovine retinal pericytes. Changes in the contractile tone were quantified as the changes in the summed length of wrinkles induced by pericytes on the silicone surface on which
H Fuder et al.
Naunyn-Schmiedeberg's archives of pharmacology, 345(4), 417-423 (1992-04-01)
To investigate whether endogenous purinoceptor agonists affect the sympathetic neurotransmission in the rat isolated iris, and to classify the purinoceptors modulating exocytotic [3H]-noradrenaline release, we have determined the effect of adenosine receptor antagonists on, and the relative potency of selected
K A Kirkpatrick et al.
British journal of pharmacology, 110(3), 949-954 (1993-11-01)
1. The effects of A1 and A2a adenosine receptor agonists on the veratridine-evoked release of [3H]-acetylcholine ([3H]-ACh) from rat striatal synaptosomes was investigated by use of the A1-selective agonist, R-PIA and the 185 fold selective A2a agonist, CGS 21680. The

Articles

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

We offers many products related to adenosine receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service