Skip to Content
Merck
All Photos(1)

Key Documents

PZ0118

Sigma-Aldrich

Nafoxidine hydrochloride

≥98% (HPLC)

Synonym(s):

1-[2-[4-(3,4-dihydro-6-methoxy-2-phenyl-1-naphthalenyl)phenoxy]ethyl]-pyrrolidine hydrochloride, CP-5600, Nafoxidene hydrochloride, PNU-0011100

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H31NO2 · HCl
CAS Number:
Molecular Weight:
462.02
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: ≥8 mg/mL

storage temp.

room temp

SMILES string

Cl.COc1ccc2c(CCC(c3ccccc3)=C2c4ccc(OCCN5CCCC5)cc4)c1

InChI

1S/C29H31NO2.ClH/c1-31-26-14-16-28-24(21-26)11-15-27(22-7-3-2-4-8-22)29(28)23-9-12-25(13-10-23)32-20-19-30-17-5-6-18-30;/h2-4,7-10,12-14,16,21H,5-6,11,15,17-20H2,1H3;1H

InChI key

HJOOGTROABIIIU-UHFFFAOYSA-N

Biochem/physiol Actions

Nafoxidine is a potent estrogen receptor antagonist that exhibits anti-proliferative properties. It is a non-steroidal antiestrogen, a derivative of Tamoxifen.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M S Wisel et al.
International journal of fertility and menopausal studies, 39(3), 156-163 (1994-05-01)
To compare effects of various anti-estrogens on early pregnancy. Fifty regularly cycling guinea pigs were divided into five groups (five animals/group), of which three groups received 3 mg/kg body weight Nafoxidine, Centchroman, or Tamoxifen, respectively, on the 1st, 2nd, or
A Yazdanyar et al.
Journal of biomedical science, 8(2), 153-159 (2001-04-05)
Microbial virulence is generally considered to be multifactorial with infection resulting from the sum of several globally regulated virulence factors. Estrogen may serve as a signal for global virulence induction in Candida albicans. Nonsteroidal estrogens and estrogen receptor antagonists may
J W Liu et al.
Biological chemistry, 380(11), 1341-1345 (1999-12-30)
The budding yeast Saccharomyces cerevisiae has been used extensively as a biological 'test tube' to study the regulation of the human estrogen receptor (ER) alpha. However, anti-estrogens, which are of great importance as therapeutic agents and research tools, fail to
Tamar Hayon et al.
Cancer detection and prevention, 27(5), 389-396 (2003-10-31)
The antileukemic activity of nonsteroidal antiestrogens was investigated. Tamoxifen, clomiphene and nafoxidine caused a decrease in viability of the estrogen receptor-negative T-lymphoblastic leukemia cell line CCRF/CEM, nafoxidine being the most active. A combination of clomiphene and genistein resulted in a
M Nichols et al.
Molecular endocrinology (Baltimore, Md.), 11(7), 950-961 (1997-06-01)
The ligand-binding domains of steroid receptors convey ligand-dependent regulation to certain proteins to which they are fused. Here we characterize fusion proteins between a site-specific recombinase, FLP, and steroid receptor ligand-binding domains. These proteins convert ligand binding into DNA recombination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service