Skip to Content
Merck
All Photos(3)

Key Documents

PLA0096

Sigma-Aldrich

Rabbit anti-p300 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

300kD, E1A-associated protein p300, E1A-binding protein, KAT3B, RSTS2, histone acetyltransferase p300, p300, p300 HAT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human

technique(s)

ChIP: 10 μg
immunohistochemistry: 1:1,000- 1:5,000
immunoprecipitation (IP): 1-5 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_001420.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... p300(2033)

Immunogen

The epitope recognized by PLA0096 maps to a region between residues 950 and 1000 of human E1A-associated protein p300 using the numbering given in entry NP_001420.2 (GeneID 2033).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

By sequence similarity p300 is related to CBP (CREB-binding protein) and functions as a histone acetyltransferase that can regulate transcription by influencing chromatin organization. P300 has been demonstrated to bind to phosphorylated CREB to mediate cAMP-gene regulation. P300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service