Skip to Content
Merck
All Photos(10)

Key Documents

ZRB1204

Sigma-Aldrich

Anti-p300 Antibody, clone 4E5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4E5, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 264.16 kDa
observed mol wt ~280 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4E5 a ZooMAb® Rabbit recombinant monoclonal antibody that detects Histone acetyltransferase p300. It targets an epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 233 amino acids from the N-terminal half of human Histone acetyltransferase p300.

Application

Quality Control Testing

Evaluated by Immunocytochemistry in MCF-7 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected p300 in MCF-7 cells.

Tested applications

ELISA Analysis: 300 μg/mL from a representative lot detected p300 in an ELISA.

Affinity Binding Assay: A representative lot of this antibody bound p300 with a KD of 2 x 10-7 in an affinity binding assay.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected p300 in nuclear extracts from HeLa and NTERA2 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-p300, clone 4E5 ZooMAb®, Cat. No. ZRB1204, is a recombinant Rabbit monoclonal antibody that detects p300 (Histone Acetyltransferase p300) and is tested for use Affinity Binding Assay, ELISA, Immunocytochemistry, and Western Blotting.

Target description

Histone acetyltransferase p300 (UniProt: Q09472; also known as EC:2.3.1.48, p300 HAT, E1A-associated protein p300, Histone butyryltransferase p300, Histone crotonyltransferase p300, Protein 2-hydroxyisobutyryltransferase p300, Protein propionyltransferase p300) is encoded by the EP300 (also known as P300) gene (Gene ID: 2033) in human. p300 serves as a transcriptional co-activator protein that plays a central role in coordinating and integrating multiple signal-dependent transcriptional events, such as cell proliferation, differentiation, and apoptosis. It has histone acetyltransferase (HAT) activity that transfers an acetyl group to the -amino group of multiple lysine residues. p300 localizes to active chromatin and acetylates all four core histones in nucleosomes, providing an epigenetic tag for transcriptional activation. It also shown to mediate cAMP-gene regulation by binding specifically to phosphorylated CREB protein. It mediates acetylation of histone H3 at lysine 122, a modification that localizes at the surface of the histone octamer and stimulates transcription, possibly by promoting nucleosome instability. It also mediates acetylation of histone H3 at lysine 27. It can also acetylase a number of non-histone targets, such as ALX1, HDAC1, PRMT1 or SIRT2. Its acetylation of HDAC1 is shown to lead to inactivation and modulation of transcription. p300 contains one TAZ-type 1 (aa 331-417), one TAZ-type 2 (aa 1728-1809), and one ZZ-type (aa 1664-1707) zinc finger domains. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service