Skip to Content
Merck
All Photos(1)

Documents

PIC0007

Sigma-Aldrich

Protease Inhibitor Cocktail

ReadyShield® ready-to-use solution, for the inhibition of serine, cysteine, aspartic and metalloproteases, Safe for Proteomic Mass Spectrometry applications

Synonym(s):

Protease inhibitor cocktail, mass spectrometry protease inhibitor cocktail

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

product name

ReadyShield® Protease Inhibitor Cocktail, Safe for Proteomic Mass Spectrometry applications

Quality Level

form

ready-to-use solution

shipped in

wet ice

storage temp.

−20°C

General description

This ReadyShield® Protease Inhibitor Cocktail is designed to be safe for mass spectrometry proteomic analysis. The protease inhibitors in the cocktail have broad specificity for the inhibition of serine, cysteine, aspartic, and metalloproteases.

This cocktail is supplied as a ready-to-use solution using a proprietary non-freezing formulation

Features and Benefits

  • Does not freeze in storage for quick, convenient use.
  • The inhibitors were selected to have no interference with LC-MS analyses.
  • Re-closable screw-top vials.
  • Can be stored in cooler for approximately 1 month or room temperature for a week.

Preparation Note

Suggested as 1000X solution.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

ReadyShield is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

ReadyShield® is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service