Skip to Content
Merck

PIC0002

Protease Inhibitor Cocktail

ReadyShield® ready-to-use solution, for the inhibition of serine, cysteine, acid proteases and aminopeptidases, for use with mammalian cell and tissue extracts, Non-freezing solution

Synonym(s):

Protease inhibitor cocktail, general use protease inhibitor cocktail

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54
Form:
ready-to-use solution
Solubility:
water: soluble
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

ReadyShield® Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, Non-freezing solution

description

Mammalian Cell and Tissue Extracts

Quality Level

form

ready-to-use solution

quality

200

packaging

pkg of 1 mL (in amber polypropylene vial), pkg of 5 mL (in amber polypropylene vial)

solubility

water: soluble

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This protease inhibitor cocktail has been optimized and tested for mammalian cell and tissue extracts. It contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases. The cocktail has been tested on extracts from various animal tissues such as human placenta and bovine liver, and on extracts of A431, CHO, and U937 cells.

This cocktail is supplied as a ready-to-use solution using a proprietary non-freezing formulation.

ReadyShield PIC0002 is a non-freezing formulation that contains the same inhibitors as is the DMSO based cocktail P8340.

Features and Benefits

  • Does not freeze in storage for quick, convenient use.
  • Tailored for Mammalian cell and tissue extracts.
  • Suggested as 100 X solution.
  • Reclosable screw-top vials.
  • Can be stored in cooler for approx. 1 month or room temperature for a week.

Preparation Note

The recommended dilution of the cocktail in the biological extract is 1 ml of the cocktail in 100 ml of lysate derived for example from 20g (wet weight) of bovine liver or 109 CHO cells

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

ReadyShield is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
PIC0002-1ML04061841255130
PIC0002-5ML04061841255147

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service