Skip to Content
Merck
All Photos(1)

Key Documents

PENNA

Sigma-Aldrich

Penicillin G sodium salt

~1650 U/mg

Synonym(s):

Benzylpenicillin sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N2NaO4S
CAS Number:
Molecular Weight:
356.37
Beilstein:
3834217
EC Number:
MDL number:
UNSPSC Code:
51283424
PubChem Substance ID:
NACRES:
NA.85

form

powder

specific activity

~1650 U/mg

solubility

H2O: 100 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)Cc3ccccc3)C([O-])=O

InChI

1S/C16H18N2O4S.Na/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1

InChI key

FCPVYOBCFFNJFS-LQDWTQKMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Penicillin G is a narrow spectrum antibiotic. It is effective against Streptococcus pneumoniae, groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridans group streptococci, and non-penicillinase producing staphylococcus. Penicillin G contains β-lactam ring in their chemical structure.

Application

Penicillin G sodium salt has been used:
  • to study the diagnostic and therapeutic implications of gentamicin-resistant Enterococcus faecalis sequence type 6 with reduced penicillin susceptibility
  • in cell culture alone as well as in combination with streptomycin and other antibiotics
  • to inhibit the synthesis of bacterial cell walls by inhibition of the cell wall peptidoglycan chain cross-lining

Biochem/physiol Actions

Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking.

Antimicrobial spectrum: This product is active against gram-positive and gram-negative bacteria.

Packaging

1mu,10mu,100mu

Caution

Solutions should be filter sterilized and stored at 2-8°C for 1 week or at -20°C for more lengthy periods. Solutions are stable at 37°C for 3 days. The sodium salt is soluble in H2O at 100 mg/mL.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.Product contains Penicillin

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J P Salanitro et al.
Applied and environmental microbiology, 32(4), 623-632 (1976-10-01)
Morphological and physiological studies were made on chicken cecal isolates of the strictly anaerobic bacterial species Gemmiger formicilis. Structural features (phase-contrast and electron microscopy) of these microorganisms indicate they (i) are highly pleomorphic, (ii) possess a trilaminar cell wall like
P Giesbrecht et al.
Journal of bacteriology, 174(7), 2241-2252 (1992-04-01)
Electron microscopic research into the murosomes of staphylococci has shown that the number of murosomes involved in penicillin-induced death varies depending on the experimental conditions employed. With 0.1 micrograms of penicillin G per ml, only 1 of a total of
Xizhen Jiang et al.
European journal of medicinal chemistry, 46(8), 3526-3530 (2011-05-17)
Thirteen compounds, based on benzofuran skeleton bearing aryl substituents at its C-3 position through methanone linker, were synthesized and screened for their antibacterial and antifungal activities against four bacteria Escherichia coli, Staphylococcus aureus, Methicillin-resistant S.aureus, Bacillus subtilis, and a fungus
Paul P Drury et al.
Neuropharmacology, 83, 62-70 (2014-04-15)
Basal ganglia injury after hypoxia-ischemia remains common in preterm infants, and is closely associated with later cerebral palsy. In the present study we tested the hypothesis that a highly selective neuronal nitric oxide synthase (nNOS) inhibitor, JI-10, would improve survival
Olivier Marion et al.
Bioorganic & medicinal chemistry, 17(3), 1006-1017 (2008-02-01)
A series of simplified thiomarinol derivatives was prepared by way of catalytic enantioselective inverse electron demand hetero [4+2] cycloaddition/allylboration tandem reaction. As a preliminary evaluation, these analogs were tested for antimicrobial activity using a standard disk diffusion assay. Whereas amide

Articles

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

β-lactam antibacterials inhibit transpeptidase enzymes, preventing peptidoglycan assembly in both Gram-positive and Gram-negative bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service