Skip to Content
Merck
All Photos(2)

Key Documents

P8287

Sigma-Aldrich

o-Phenylenediamine dihydrochloride

chromogenic, tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3912045
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

o-Phenylenediamine dihydrochloride, tablet, 10 mg substrate per tablet

form

tablet

mp

258 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Phenylenediamine (o-PD) is a colorless and nonfluorescent molecule.
o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

o-Phenylenediamine dihydrochloride has been used as a substrate for horseradish-peroxidase-conjugated secondary antibody in enzyme-linked immunosorbent assay (ELISA).

Reconstitution

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara Buezo Montero et al.
Frontiers in cellular and infection microbiology, 10, 377-377 (2020-08-28)
Mosquitoes of the Aedes genus transmit arboviruses of great importance to human health as dengue, chikungunya, Zika and yellow fever. The tiger mosquito Aedes albopictus can play an important role as arboviral vector, especially when Aedes aegypti is absent or
Henry Surendra et al.
Malaria journal, 18(1), 227-227 (2019-07-10)
Analysis of anti-malarial antibody responses has the potential to improve characterization of the variation in exposure to infection in low transmission settings, where conventional measures, such as entomological estimates and parasitaemia point prevalence become less sensitive and expensive to measure.
Rama Devudu Puligedda et al.
Antibodies (Basel, Switzerland), 9(1) (2020-03-04)
Poliovirus (PV)-specific intestinal IgAs are important for cessation of PV shedding in the gastrointestinal tract following an acute infection with wild type or vaccine-derived PV strains. We sought to produce IgA monoclonal antibodies (mAbs) with PV neutralizing activity. We first
Rafaella P M Guimarães-Peixoto et al.
Veterinary parasitology, regional studies and reports, 20, 100375-100375 (2020-05-26)
Bovine cysticercosis remains as one of the most important cause of carcasses and viscera condemnation in Brazilian slaughterhouses. However, the efficiency of post-mortem inspection for the diagnosis of this zoonotic disease is relatively low, and few available studies were performed
Saima Ajaz et al.
American journal of physiology. Cell physiology, 320(1), C57-C65 (2020-11-06)
The COVID-19 pandemic has been the primary global health issue since its outbreak in December 2019. Patients with metabolic syndrome suffer from severe complications and a higher mortality rate due to severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection. We

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service