Skip to Content
Merck

Skip To

P1063

o-Phenylenediamine dihydrochloride

tablet, 60 mg substrate per tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
UNSPSC Code:
12352204
NACRES:
NA.83
PubChem Substance ID:
EC Number:
210-418-7
Beilstein/REAXYS Number:
3912045
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

tablet

technique(s)

protein detection: suitable

mp

258 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

General description

o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.
o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

o-Phenylenediamine dihydrochloride has been used as a stop solution for evaluating bovine serum albumin (BSA) antibodies via enzyme-linked immunosorbent assay (ELISA). It has also been used to measure specific antibodies in pig serum by ELISA.

Features and Benefits

Supplied as 50 or 100 tablets per box, individually wrapped with foil for easy storage, use, and safety.

Preparation Note

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P8806P7288P8412
description

tablet, 60 mg substrate per tablet

description

tablet, 3 mg substrate per tablet

description

tablet, 20 mg substrate per tablet

description

tablet, 30 mg substrate per tablet

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

tablet

form

tablet

form

tablet

form

tablet

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

mp

258 °C (dec.) (lit.)

mp

258 °C (dec.) (lit.)

mp

258 °C (dec.) (lit.)

mp

258 °C (dec.) (lit.)


Still not finding the right product?

Explore all of our products under o-Phenylenediamine dihydrochloride


Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters