Skip to Content
Merck
All Photos(1)

Key Documents

P5400

Sigma-Aldrich

Pectinesterase from orange peel

greener alternative

lyophilized powder, ≥150 units/mg protein

Synonym(s):

Pectin methylesterase, Pectin pectylhydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

specific activity

≥150 units/mg protein

composition

Protein, 20-50% biuret

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been used as enzyme for alternative energy. For more information see the Enzymes for Alternative Energy Research.

Application

Pectinesterase is used to hydrolyze methyl esters of pectin to pectate and methanol. It contains pectin methylesterase and pectin pectylhydrolase. Product P5400 is from orange peel and comes as a lyophilized powder. It contains (NH4)2SO4 and sodium chloride. It has been used to study the pectin methylesterase gene.
The enzyme from Sigma has been used as a standard during the measurement of pectin methylesterase activity in sea buckthorn juices. It has been used in the hydrolysis of cell wall pectins while studying phyllotaxis in Arabidopsis. It has also been used to catalyze pectin hydrolysis (using an ultrasonic measurement device) for monitoring biological processes.

Biochem/physiol Actions

Pectinesterase catalyzes the hydrolysis of the methyl esters of pectin to yield pectate and methanol.

Unit Definition

One unit will release 1.0 microequivalent of acid from pectin per min at pH 7.5 at 30 °C.

Physical form

Contains (NH4)2SO4 and sodium chloride

Analysis Note

Protein determined by biuret.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuri L Dorokhov et al.
PLoS pathogens, 8(4), e1002640-e1002640 (2012-04-13)
Many plants release airborne volatile compounds in response to wounding due to pathogenic assault. These compounds serve as plant defenses and are involved in plant signaling. Here, we study the effects of pectin methylesterase (PME)-generated methanol release from wounded plants
Daniele Del Corpo et al.
Molecular plant pathology, 21(12), 1620-1633 (2020-10-09)
Pectin is synthesized in a highly methylesterified form in the Golgi cisternae and partially de-methylesterified in muro by pectin methylesterases (PMEs). Arabidopsis thaliana produces a local and strong induction of PME activity during the infection of the necrotrophic fungus Botrytis
Kerstin Müller et al.
Plant physiology, 161(1), 305-316 (2012-11-07)
The methylesterification status of cell wall homogalacturonans, mediated through the action of pectin methylesterases (PMEs), influences the biophysical properties of plant cell walls such as elasticity and porosity, important parameters for cell elongation and water uptake. The completion of seed
Mélanie L'Enfant et al.
International journal of biological macromolecules, 132, 1140-1146 (2019-04-13)
The discovery of molecules that can inhibit the action of phytopathogens is essential to find alternative to current pesticides. Pectin methylesterases (PME), enzymes that fine-tune the degree of methylesterification of plant cell wall pectins, play a key role in the
Ida Barbara Reca et al.
Plant molecular biology, 79(4-5), 429-442 (2012-05-23)
A pectin methylesterase inhibitor (SolyPMEI) from tomato has been identified and characterised by a functional genomics approach. SolyPMEI is a cell wall protein sharing high similarity with Actinidia deliciosa PMEI (AdPMEI), the best characterised inhibitor from kiwi. It typically affects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service