Skip to Content
Merck
All Photos(2)

Key Documents

P2401

Sigma-Aldrich

Pectinase from Rhizopus sp.

greener alternative

powder, 400-800 units/g solid

Synonym(s):

Macerozyme R-10, Poly-(1,4-α-D-galacturonide) glycanohydrolase, Polygalacturonase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

specific activity

400-800 units/g solid

greener alternative product characteristics

Waste Prevention
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and waste prevention when used in cellulosic ethanol research. For more information see the article in biofiles and Enzymes for Alternative Energy Research.

Pectinase, also known as pectinolytic enzyme, is predominantly found in bacteria, fungi and plants. Polygalacturonase is one of the highly abundant enzymes among all the pectinolytic enzymes. Pectinase has a wide range of industrial applications including, fruit juice extraction and its clarification. In addition, it is also used in vegetable oil extraction, tea and coffee fermentations, bleaching of paper, in poultry feed additives and in alcoholic beverages and food industries.

Application

Pectinase from Rhizopus sp has been used for enzymatic digestion of leaves.
Used in plant protoplast preparation to digest cell wall prior to organelle isolation.

Biochem/physiol Actions

Pectinases hydrolyses pectin, which is a component of the cell wall.They may attack methyl-esterified pectin or de-esterified pectin. It is a source of pectinase activity, also containing cellulase and hemicellulase activities.

Unit Definition

One unit will liberate 1.0 μmole of galacturonic acid from polygalacturonic acid per min at pH 4.0 at 25 °C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pectinase production by Neurospora crassa: purification and biochemical characterization of extracellular polygalacturonase activity
Maria de Lourdes T. M. Polizeli, et al.
Microbiology, 137, 1815-1823 (1991)
Sheetal K Karnik et al.
Plant physiology, 138(4), 1967-1981 (2005-07-26)
Homologs of peroxin 16 genes (PEX16) have been identified only in Yarrowia lipolytica, humans (Homo sapiens), and Arabidopsis (Arabidopsis thaliana). The Arabidopsis gene (AtPEX16), previously reported as the SSE1 gene, codes for a predicted 42-kD membrane peroxin protein (AtPex16p). Lin
Isolation of intact sub-dermal secretory cavities from Eucalyptus
Goodger JQ, et al.
Plant methods, 6(1), 20-20 (2010)
Enzyme disintegration with spatial resolution reveals different distributions of sludge extracellular polymer substances
Lu Fan, et al.
Biotechnology for Biofuels, 9(1), 29-29 (2016)
Microbial pectinolytic enzymes: a review
Jayani Ranveer Singh, et al.
Process. Biochem., 40(9), 2931-2944 (2005)

Protocols

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

To measure pectinase activity, a titrimetric stop reaction assay is used. One unit of pectinase will liberate 1 μmol of galacturonic acid from poly-galacturonic acid per hour at pH 4.0 at 25 °C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service