Skip to Content
Merck
All Photos(1)

Documents

P2143

Sigma-Aldrich

Protease from Aspergillus saitoi

Type XIII, ≥0.6 unit/mg solid

Synonym(s):

Molsin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

biological source

Aspergillus sp. (Aspergillus saitoi)

type

Type XIII

form

solid

specific activity

≥0.6 unit/mg solid

foreign activity

alkaline protease, essentially free

storage temp.

−20°C

Application

Protease from Aspergillus saitoi has been used in a study to assess sequence coverage and resolution in hydrogen exchange of large proteins. It has also been used in a study to investigate the conversion of soybean curd isoflavone glycosides to their aglycones through β-glucosidase..

Biochem/physiol Actions

Protease from Aspergillus saitoi was shown to also function as a β-glucosidase.

Unit Definition

One unit will hydrolyze hemoglobin to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 2.8 at 37 °C (color by Folin-Ciocalteu reagent).

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Moralejo et al.
Applied microbiology and biotechnology, 54(6), 772-777 (2001-01-11)
A gene encoding the sweet-tasting protein thaumatin (tha) with optimized codon usage was expressed in Aspergillus awamori. Mutants of A. awamori with reduced proteolytic activity were isolated. One of these mutants, named lpr66, contained an insertion of about 200 bp
Mizuho Nishinoaki et al.
Bioscience, biotechnology, and biochemistry, 72(2), 587-590 (2008-02-08)
An Aspergillus saitoi protease preparation, Molsin, was found to contain beta-glucosidase as well as protease activities. Application of Molsin to soybean curd improved its functionality by converting the contained isoflavone glycosides to their aglycones through beta-glucosidase, and also modified the
E Ichishima
Bioscience, biotechnology, and biochemistry, 64(4), 675-688 (2000-06-01)
This review covers the unique catalytic and molecular properties of three proteolytic enzymes and a glycosidase from Aspergillus. An aspartic proteinase from A. saitoi, aspergillopepsin I (EC 3.4.23.18), favors hydrophobic amino acids at P1 and P'1 like gastric pepsin. However
M E Kambouris et al.
FEMS immunology and medical microbiology, 25(3), 255-264 (1999-08-25)
In immunodeficient patients, Aspergillus species emerge as circumstantial pathogens. Aspergillus fumigatus is a distant first among the pathogenic aspergilli, which cause deep-seated mycoses. Sequences of the pep gene of A. fumigatus as potential PCR primers, which have not been tested
S W Cho et al.
Acta crystallographica. Section D, Biological crystallography, 57(Pt 7), 948-956 (2001-06-22)
The crystal structure of aspergillopepsin I (AP) from Aspergillus phoenicis has been determined at 2.18 A resolution and refined to R and R(free) factors of 21.5 and 26.0%, respectively. AP has the typical two beta-barrel domain structure of aspartic proteinases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service