P6110
Protease from Aspergillus oryzae
≥500 U/g
Synonym(s):
Flavourzyme®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
liquid
specific activity
≥500 U/g
storage temp.
2-8°C
Related Categories
General description
Fungal protease/peptidase complex produced by submerged fermentation of a selected strain of Aspergillus oryzae and contains both endoprotease and exopeptidase activities.
Proteases belong to the group of hydrolases and exist as acid, neutral, and alkaline proteases.
Application
Protease from Aspergillus oryzae has been used:
- to study the effect of protease inhibitors from Moringa oleifera
- to evaluate its hydrolyzing effect on the anatomical structure and chemical composition of archaeological wood samples
- to perform protease digestion to process mucosa samples obtained from the colon of pigs
Protease is used to degrade proteins, to study protease inhibitors and to study thermal inactivation kinetics. Protease is used in nucleic acid isolation procedures in incubations. It has been injected into flies with a nanoject apparatus for infection and survival experiments.
The enzyme from Sigma has been used in the semi-purification of mouse colorectal mucins during protein digestion.
Biochem/physiol Actions
Proteases catabolize proteins by hydrolysis of peptide bonds. They have many applications such as in detergents, bioremediation processes, pharmaceutical industry, and food industries. Proteases are associated with nitrogen mineralization in the soil. They also serve as a supplement in swine and poultry diets.
Unit Definition
One unit is the amount of enzyme which hydrolyzes 1 mmol of L-Ieucine-p-nitroanilide per minute.
Legal Information
A product of Novozyme Corp.
Flavourzyme is a registered trademark of Novozymes Corp.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hydrolases of Halophilic Origin With Importance for the Food Industry
Enzymes in Food Biotechnology, 93-109 (2019)
Jelena Patrnogic et al.
PloS one, 12(12), e0188339-e0188339 (2017-12-07)
In Drosophila, recognition of pathogens such as Gram-positive bacteria and fungi triggers the activation of proteolytic cascades and the subsequent activation of the Toll pathway. This response can be achieved by either detection of pathogen associated molecular patterns or by
Sunantha Ketnawa et al.
Chemistry Central journal, 7(1), 79-79 (2013-05-02)
Too many different protein and enzyme purification techniques have been reported, especially, chromatographic techniques. Apart from low recovery, these multi-step methods are complicated, time consuming, high operating cost. So, alternative beneficially methods are still required. Since, the outstanding advantages of
Refined methodology to purify mucins from pig colonic mucosa
Libao-Mercado AJ and De Lange CFM
Livestock Science, 109(1-3), 141-144 (2007)
Exploiting microbial enzymes for augmenting crop production
Enzymes in Food Biotechnology, 503-519 (2019)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service