Skip to Content
Merck
All Photos(2)

Documents

P1178

Sigma-Aldrich

Phloroglucinol

suitable for plant cell culture, BioReagent

Synonym(s):

1,3,5-Trihydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
1341907
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72

product line

BioReagent

Quality Level

form

powder

technique(s)

cell culture | plant: suitable

application(s)

agriculture

SMILES string

Oc1cc(O)cc(O)c1

InChI

1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H

InChI key

QCDYQQDYXPDABM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phloroglucinol is a trihydroxybenzene with antithrombotic, profibrinolytic, antimicrobial, antimalarial, cancer chemopreventive, anti-HIV and anti-leishmanial activities. Phloroglucinol (PG) is a biosynthetic precursor of the 2,4-diacetylphloroglucinol (DAPG) an antibiotic against soil-borne diseases. Phloroglucinol is a useful intermediate because it is polyfunctional.

Application

Phloroglucinol has been used to prevent hyperhydricity by promoting lignification of shoots. It has also been used in PG (phloroglucinol)-sucrose treatments for growth and development of in vitro derived shoot tips.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phloroglucinol enhances growth and rate of axillary shoot proliferation in potato shoot tip cultures in vitro
Sarkar D and Naik P S
Plant Cell, Tissue and Organ Culture, 60(2), 139-149 (2000)
In vitro propagation of `Guayabo del pais?(Acca sellowiana (Berg.) Burret)
Ross S and Grasso R
Fruit, Vegetable and Cereal Science and Biotechnology, 4(1), 83-87 (2010)
Jong-Sup Bae
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 49(7), 1572-1577 (2011-04-20)
Phloroglucinol is the monomeric units of phlorotannins abundant in brown algae. Several biological effects of phloroglucinol have been reported, however, antithrombotic and profibrinolytic activities of phloroglucinol have not been studied yet. In this study, the anticoagulant properties of phloroglucinol were
Yong Li et al.
Bioorganic & medicinal chemistry, 17(5), 1963-1973 (2009-02-10)
Seven phlorotannins were isolated and characterized from an edible marine brown alga Ecklonia cava (EC), along with three common sterol derivatives (fucosterol, ergosterol, and cholesterol) according to the comprehensive spectral analysis of MS and NMR data. Compounds 5 (7-phloro eckol)
Liva Harinantenaina et al.
Journal of natural products, 76(3), 388-393 (2013-01-05)
Bioassay-guided fractionation of an ethanol extract of the leaves and inflorescence of Mallotus oppositifolius collected in Madagascar led to the isolation of the two new bioactive dimeric phloroglucinols mallotojaponins B (1) and C (2), together with the known mallotophenone (3).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service