Skip to Content
Merck
All Photos(5)

Key Documents

P1138

Sigma-Aldrich

1,2-Distearoyl-sn-glycero-3-phosphocholine

≥99%

Synonym(s):

PC, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, 1,2-distearoyl, L-α-Phosphatidylcholine, distearoyl, L-β,γ-Distearoyl-α-lecithin, DSPC, PC(18:0/18:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H88NO8P
CAS Number:
Molecular Weight:
790.15
Beilstein:
3923978
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

semisynthetic

Quality Level

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1

InChI key

NRJAVPSFFCBXDT-HUESYALOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Non-pyrogenic, well-defined liposomes, loaded with a molecule of choice, are formed by a single hydration step.

Application


  • Amantadine interactions with phase separated lipid membranes.: This study explores the interactions between amantadine and phase-separated lipid membranes, providing insights into the role of 1,2-Distearoyl-sn-glycero-3-phosphocholine in membrane structure and function (Kinnun et al., 2024).

  • Design of charge converting lipid nanoparticles via a microfluidic coating technique.: This research designs lipid nanoparticles with charge-converting capabilities using 1,2-Distearoyl-sn-glycero-3-phosphocholine, enhancing drug delivery systems (Zöller et al., 2024).

  • Investigation and Comparison of Active and Passive Encapsulation Methods for Loading Proteins into Liposomes.: The study compares methods for protein encapsulation into liposomes using 1,2-Distearoyl-sn-glycero-3-phosphocholine, advancing drug delivery technologies (Pisani et al., 2023).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter J Kueffer et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(16), 6512-6517 (2013-03-29)
The application of boron neutron capture therapy (BNCT) following liposomal delivery of a (10)B-enriched polyhedral borane and a carborane against mouse mammary adenocarcinoma solid tumors was investigated. Unilamellar liposomes with a mean diameter of 134 nm or less, composed of
Sweta Modi et al.
Journal of controlled release : official journal of the Controlled Release Society, 162(2), 330-339 (2012-07-18)
Nanoparticulate drug carriers such as liposomal drug delivery systems are of considerable interest in cancer therapy because of their ability to passively accumulate in solid tumors. For liposomes to have practical utility for antitumor therapy in patients, however, optimization of
Pan Li et al.
Journal of controlled release : official journal of the Controlled Release Society, 162(2), 349-354 (2012-07-18)
Ultrasound targeted microbubble destruction (UTMD) was one of the most promising strategies to enhance drug delivery in cancer therapy. Microbubbles (MBs) serve as a vehicle to carry anti-tumor drugs and locally release them when exposed to therapeutic ultrasound, resulting in
Mette Marie Bruun Nielsen et al.
Langmuir : the ACS journal of surfaces and colloids, 29(5), 1525-1532 (2013-01-04)
Imaging ellipsometry (IE) has been applied to generate laterally resolved thickness maps of spin-coated membranes in both the dry and fully hydrated state. Spin-coating offers a convenient preparation method for stacked supported membranes, and in-depth thickness maps for such films
Hiroyuki Nakamura et al.
Organic & biomolecular chemistry, 10(7), 1374-1380 (2011-12-24)
The fluorescence-labeled closo-dodecaborane lipid (FL-SBL) was synthesized from (S)-(+)-1,2-isopropylideneglycerol as a chiral starting material. FL-SBL was readily accumulated into the PEGylated DSPC liposomes prepared from DSPC, CH, and DSPE-PEG-OMe by the post insertion protocol. The boron concentrations and the fluorescent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service