Skip to Content
Merck
All Photos(1)

Key Documents

P0300

Sigma-Aldrich

Protein Kinase Inhibitor from rabbit

≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C94H148N32O31
CAS Number:
Molecular Weight:
2222.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

rabbit

Assay

≥85% (HPLC)

form

powder

mol wt

2.2 kDa

composition

Peptide content, ≥70%

solubility

water: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(C)NC(=O)C(CC(N)=O)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCNC(N)=N)NC(=O)CNC(=O)C(NC(=O)C(CCCNC(N)=N)NC(=O)CNC(=O)C(CO)NC(=O)C(C)NC(=O)C(NC(=O)C(Cc1ccccc1)NC(=O)C(CC(O)=O)NC(=O)C(C)NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(NC(=O)C(N)C(C)O)C(C)O)C(C)CC)C(C)O)C(=O)NC(Cc3cnc[nH]3)C(=O)NC(CC(O)=O)C(O)=O

InChI

1S/C94H148N32O31/c1-11-42(3)70(124-85(150)58(31-50-19-14-13-15-20-50)117-84(149)61(35-67(135)136)116-74(139)44(5)110-81(146)57(32-51-24-26-53(131)27-25-51)119-90(155)73(49(10)130)126-86(151)69(96)47(8)128)88(153)112-45(6)75(140)122-63(40-127)77(142)107-38-65(133)114-55(22-17-29-105-93(99)100)80(145)125-72(48(9)129)87(152)108-39-66(134)113-54(21-16-28-104-92(97)98)78(143)115-56(23-18-30-106-94(101)102)79(144)118-60(34-64(95)132)82(147)111-46(7)76(141)123-71(43(4)12-2)89(154)120-59(33-52-37-103-41-109-52)83(148)121-62(91(156)157)36-68(137)138/h13-15,19-20,24-27,37,41-49,54-63,69-73,127-131H,11-12,16-18,21-23,28-36,38-40,96H2,1-10H3,(H2,95,132)(H,103,109)(H,107,142)(H,108,152)(H,110,146)(H,111,147)(H,112,153)(H,113,134)(H,114,133)(H,115,143)(H,116,139)(H,117,149)(H,118,144)(H,119,155)(H,120,154)(H,121,148)(H,122,140)(H,123,141)(H,124,150)(H,125,145)(H,126,151)(H,135,136)(H,137,138)(H,156,157)(H4,97,98,104)(H4,99,100,105)(H4,101,102,106)

InChI key

AXOXZJJMUVSZQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp

Biochem/physiol Actions

Inhibitor of c-AMP-dependent protein kinase
These proteins bind to the catalytic subunit of cyclic-AMP dependent protein kinase, displacing the regulatory subunit and inhibiting phosphorylating activity. The cyclic-AMP binding capacity of the regulatory subunit is enhanced.

Features and Benefits

This compound is featured on the PKA & PKG page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M V Autieri et al.
Cell regulation, 1(13), 1015-1025 (1990-12-01)
We have shown previously that cytoplasmic extracts from actively dividing lymphoid cells are capable of inducing DNA synthesis in isolated nuclei. One of the factors involved in this activity, ADR, appears to be a greater than 90 kDa heat-labile protease.
Z Wang et al.
The Journal of biological chemistry, 273(1), 73-79 (1998-02-07)
Cadmium is mitogenic under some circumstances and has been shown to cause accumulation of transcripts for several proto-oncogenes in a variety of cells, but the mechanism(s) remain to be delineated. Here we show that CdCl2 causes an increase in c-fos
Li Chen et al.
Food chemistry, 293, 537-544 (2019-06-04)
To verify the effect of protein phosphorylation on glycolysis and elucidate the regulatory mechanism from the perspective of enzyme activity, ovine muscle was treated with a kinase inhibitor, dimethyl sulfoxide, or a phosphatase inhibitor and the activities of glycogen phosphorylase
T Miralem et al.
The Journal of biological chemistry, 271(29), 17100-17106 (1996-07-19)
Heparin suppresses mitogenic responses in renal mesangial cells, and when quiescent mesangial cells are stimulated with serum, heparin blocks the induction of c-fos seen at 15 min. Because heparin is taken up by cells over a much longer time course
J W Kasckow et al.
Synapse (New York, N.Y.), 18(4), 354-358 (1994-12-01)
Interleukin-1 (IL1) is a key messenger implicated in endocrine and immune systems that interact to mediate the stress response. Corticotropin-releasing factor (CRF) secretion and synthesis in the NPLC-KC human hepatoma cell line has been shown to respond to IL1 stimulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service